Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101684026-101684281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARSL2
Entrez Gene ID
123283
Gene Name
threonyl-tRNA synthetase like 2
Gene Aliases
-
UniGene
Hs.657163
Gene Chromosome Location
Chr.15: 101653752-101724604 on Build GRCh38
Transcripts
NM_152334.2 XM_006720386.3 XM_017021912.1 XM_017021913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
959714 c.1500C>A p.A500A Chr.15: 101684225-101684225 on Build GRCh38
959713 c.1524A>G p.E508E Chr.15: 101684201-101684201 on Build GRCh38
4641908 c.1614G>C p.Q538H Chr.15: 101684111-101684111 on Build GRCh38
6796903 c.1511G>A p.R504Q Chr.15: 101684214-101684214 on Build GRCh38
4053557 c.1629C>T p.H543H Chr.15: 101684096-101684096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767758561 Chr.15: 101684200-101684200 on Build GRCh38
rs755637634 Chr.15: 101684057-101684058 on Build GRCh38
rs34201431 Chr.15: 101684119-101684119 on Build GRCh38
rs765089231 Chr.15: 101684078-101684078 on Build GRCh38
rs759263507 Chr.15: 101684151-101684151 on Build GRCh38
rs982643114 Chr.15: 101684150-101684150 on Build GRCh38
rs567399004 Chr.15: 101684064-101684066 on Build GRCh38
rs758891836 Chr.15: 101684221-101684221 on Build GRCh38
rs767664849 Chr.15: 101684081-101684081 on Build GRCh38
rs775321387 Chr.15: 101684170-101684170 on Build GRCh38
rs893294662 Chr.15: 101684084-101684084 on Build GRCh38
rs779302720 Chr.15: 101684218-101684218 on Build GRCh38
rs1008577017 Chr.15: 101684197-101684197 on Build GRCh38
rs906845736 Chr.15: 101684254-101684254 on Build GRCh38
rs759583538 Chr.15: 101684067-101684067 on Build GRCh38
rs754039316 Chr.15: 101684214-101684214 on Build GRCh38
rs1031863575 Chr.15: 101684074-101684074 on Build GRCh38
rs755326278 Chr.15: 101684215-101684215 on Build GRCh38
rs761049662 Chr.15: 101684211-101684211 on Build GRCh38
rs766518014 Chr.15: 101684212-101684212 on Build GRCh38
rs1044274998 Chr.15: 101684103-101684103 on Build GRCh38
rs12901450 Chr.15: 101684138-101684138 on Build GRCh38
rs746721347 Chr.15: 101684140-101684140 on Build GRCh38
rs778758049 Chr.15: 101684232-101684233 on Build GRCh38
rs757019418 Chr.15: 101684144-101684153 on Build GRCh38
rs769584879 Chr.15: 101684161-101684161 on Build GRCh38
rs777329635 Chr.15: 101684226-101684226 on Build GRCh38
rs200095766 Chr.15: 101684145-101684145 on Build GRCh38
rs749613889 Chr.15: 101684118-101684118 on Build GRCh38
rs763685878 Chr.15: 101684067-101684067 on Build GRCh38
rs574856899 Chr.15: 101684187-101684187 on Build GRCh38
rs753465149 Chr.15: 101684095-101684096 on Build GRCh38
rs770564048 Chr.15: 101684141-101684141 on Build GRCh38
rs757307878 Chr.15: 101684080-101684080 on Build GRCh38
rs908414406 Chr.15: 101684194-101684194 on Build GRCh38
rs750716067 Chr.15: 101684085-101684085 on Build GRCh38
rs149570515 Chr.15: 101684131-101684131 on Build GRCh38
rs756387165 Chr.15: 101684104-101684104 on Build GRCh38
rs374361813 Chr.15: 101684228-101684228 on Build GRCh38
rs370695273 Chr.15: 101684137-101684137 on Build GRCh38
rs780626895 Chr.15: 101684105-101684105 on Build GRCh38
rs138665867 Chr.15: 101684251-101684251 on Build GRCh38
rs143111392 Chr.15: 101684220-101684220 on Build GRCh38
rs748646277 Chr.15: 101684133-101684133 on Build GRCh38
rs770369664 Chr.15: 101684231-101684231 on Build GRCh38
rs775335074 Chr.15: 101684256-101684256 on Build GRCh38
rs780731407 Chr.15: 101684232-101684232 on Build GRCh38
rs769518268 Chr.15: 101684252-101684252 on Build GRCh38
rs752624681 Chr.15: 101684079-101684079 on Build GRCh38
rs750520100 Chr.15: 101684201-101684201 on Build GRCh38

View Full Product Details