Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28356585-28356858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF395
Entrez Gene ID
55893
Gene Name
zinc finger protein 395
Gene Aliases
HDBP-2, HDBP2, HDRF-2, PBF, PRF-1, PRF1, Si-1-8-14
UniGene
Hs.435535
Gene Chromosome Location
Chr.8: 28345585-28386460 on Build GRCh38
Transcripts
NM_018660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825442 c.536G>A p.C179Y Chr.8: 28356717-28356717 on Build GRCh38
3899397 c.484G>A p.A162T Chr.8: 28356769-28356769 on Build GRCh38
4626215 c.558T>A p.P186P Chr.8: 28356695-28356695 on Build GRCh38
291657 c.519G>A p.V173V Chr.8: 28356734-28356734 on Build GRCh38
486367 c.558T>C p.P186P Chr.8: 28356695-28356695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199746046 Chr.8: 28356659-28356659 on Build GRCh38
rs752754037 Chr.8: 28356770-28356770 on Build GRCh38
rs903281546 Chr.8: 28356686-28356686 on Build GRCh38
rs368552072 Chr.8: 28356667-28356667 on Build GRCh38
rs372816052 Chr.8: 28356733-28356733 on Build GRCh38
rs201440297 Chr.8: 28356789-28356789 on Build GRCh38
rs771646593 Chr.8: 28356741-28356743 on Build GRCh38
rs755457105 Chr.8: 28356656-28356656 on Build GRCh38
rs2272748 Chr.8: 28356608-28356608 on Build GRCh38
rs150901935 Chr.8: 28356696-28356696 on Build GRCh38
rs933123330 Chr.8: 28356648-28356648 on Build GRCh38
rs369705922 Chr.8: 28356729-28356729 on Build GRCh38
rs758737374 Chr.8: 28356646-28356646 on Build GRCh38
rs537917726 Chr.8: 28356622-28356622 on Build GRCh38
rs370490939 Chr.8: 28356623-28356623 on Build GRCh38
rs141601441 Chr.8: 28356692-28356692 on Build GRCh38
rs568474253 Chr.8: 28356660-28356660 on Build GRCh38
rs777862697 Chr.8: 28356782-28356782 on Build GRCh38
rs781239649 Chr.8: 28356657-28356657 on Build GRCh38
rs746818066 Chr.8: 28356792-28356792 on Build GRCh38
rs536541461 Chr.8: 28356717-28356717 on Build GRCh38
rs750790071 Chr.8: 28356638-28356638 on Build GRCh38
rs946897512 Chr.8: 28356726-28356726 on Build GRCh38
rs35385862 Chr.8: 28356784-28356784 on Build GRCh38
rs768690584 Chr.8: 28356811-28356811 on Build GRCh38
rs761550424 Chr.8: 28356625-28356625 on Build GRCh38
rs778371481 Chr.8: 28356654-28356654 on Build GRCh38
rs987855535 Chr.8: 28356670-28356670 on Build GRCh38
rs772865790 Chr.8: 28356825-28356825 on Build GRCh38
rs761455144 Chr.8: 28356710-28356710 on Build GRCh38
rs776246585 Chr.8: 28356706-28356706 on Build GRCh38
rs765020463 Chr.8: 28356634-28356634 on Build GRCh38
rs748124351 Chr.8: 28356820-28356820 on Build GRCh38
rs756139270 Chr.8: 28356773-28356773 on Build GRCh38
rs769365409 Chr.8: 28356824-28356824 on Build GRCh38
rs762520973 Chr.8: 28356728-28356728 on Build GRCh38
rs760532740 Chr.8: 28356621-28356621 on Build GRCh38
rs749494663 Chr.8: 28356683-28356683 on Build GRCh38
rs772956100 Chr.8: 28356725-28356725 on Build GRCh38
rs779892847 Chr.8: 28356791-28356791 on Build GRCh38
rs781222123 Chr.8: 28356813-28356813 on Build GRCh38
rs867651999 Chr.8: 28356774-28356774 on Build GRCh38
rs752042287 Chr.8: 28356655-28356655 on Build GRCh38
rs372221668 Chr.8: 28356688-28356688 on Build GRCh38
rs376293921 Chr.8: 28356769-28356769 on Build GRCh38

View Full Product Details