Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43959731-43959964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT3
Entrez Gene ID
56894
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases
LPAAT-GAMMA1, LPAAT3
UniGene
Hs.248785
Gene Chromosome Location
Chr.21: 43865160-43987594 on Build GRCh38
Transcripts
NM_001037553.1 NM_020132.4 XM_005261160.4 XM_006724029.3 XM_006724030.3 XM_006724031.3 XM_011529660.2 XM_011529661.2 XM_011529662.2 XM_011529663.2 XM_011529664.2 XM_011529665.2 XM_017028408.1 XM_017028409.1 XM_017028410.1 XM_017028411.1 XM_017028412.1 XM_017028413.1 XM_017028414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972797 c.143G>A p.R48H Chr.21: 43959824-43959824 on Build GRCh38
6250989 c.111G>T p.A37A Chr.21: 43959792-43959792 on Build GRCh38
4660384 c.178+2T>C p.? Chr.21: 43959861-43959861 on Build GRCh38
4660383 c.110C>T p.A37V Chr.21: 43959791-43959791 on Build GRCh38
5599037 c.91G>C p.V31L Chr.21: 43959772-43959772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766368458 Chr.21: 43959870-43959870 on Build GRCh38
rs745409771 Chr.21: 43959796-43959796 on Build GRCh38
rs151002444 Chr.21: 43959800-43959800 on Build GRCh38
rs376805440 Chr.21: 43959784-43959784 on Build GRCh38
rs945497048 Chr.21: 43959926-43959926 on Build GRCh38
rs761141836 Chr.21: 43959773-43959775 on Build GRCh38
rs760861867 Chr.21: 43959840-43959840 on Build GRCh38
rs904830552 Chr.21: 43959768-43959768 on Build GRCh38
rs369874117 Chr.21: 43959791-43959791 on Build GRCh38
rs116740715 Chr.21: 43959934-43959934 on Build GRCh38
rs572753267 Chr.21: 43959778-43959778 on Build GRCh38
rs755101432 Chr.21: 43959887-43959887 on Build GRCh38
rs773426966 Chr.21: 43959771-43959772 on Build GRCh38
rs949591024 Chr.21: 43959916-43959916 on Build GRCh38
rs199595393 Chr.21: 43959818-43959818 on Build GRCh38
rs780878304 Chr.21: 43959792-43959792 on Build GRCh38
rs201739855 Chr.21: 43959835-43959835 on Build GRCh38
rs376824543 Chr.21: 43959836-43959836 on Build GRCh38
rs1050966279 Chr.21: 43959923-43959923 on Build GRCh38
rs368536410 Chr.21: 43959901-43959901 on Build GRCh38
rs903868040 Chr.21: 43959935-43959935 on Build GRCh38
rs562773406 Chr.21: 43959912-43959912 on Build GRCh38
rs1046649962 Chr.21: 43959925-43959925 on Build GRCh38
rs768508437 Chr.21: 43959814-43959814 on Build GRCh38
rs774081698 Chr.21: 43959824-43959824 on Build GRCh38
rs142811396 Chr.21: 43959772-43959772 on Build GRCh38
rs761767729 Chr.21: 43959828-43959828 on Build GRCh38
rs951134803 Chr.21: 43959799-43959799 on Build GRCh38
rs754098620 Chr.21: 43959881-43959881 on Build GRCh38
rs1028207142 Chr.21: 43959876-43959876 on Build GRCh38
rs779351025 Chr.21: 43959785-43959785 on Build GRCh38
rs73906687 Chr.21: 43959914-43959914 on Build GRCh38
rs566820643 Chr.21: 43959939-43959939 on Build GRCh38
rs191570129 Chr.21: 43959801-43959801 on Build GRCh38
rs748676164 Chr.21: 43959786-43959786 on Build GRCh38
rs753073391 Chr.21: 43959900-43959900 on Build GRCh38
rs576146849 Chr.21: 43959913-43959913 on Build GRCh38
rs765721345 Chr.21: 43959888-43959888 on Build GRCh38

View Full Product Details