Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27664245-27664500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC4A1AP
Entrez Gene ID
22950
Gene Name
solute carrier family 4 member 1 adaptor protein
Gene Aliases
HLC3
UniGene
Hs.306000
Gene Chromosome Location
Chr.2: 27663471-27694980 on Build GRCh38
Transcripts
NM_018158.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093663 c.647G>C p.R216T Chr.2: 27664399-27664399 on Build GRCh38
1407517 c.599G>A p.C200Y Chr.2: 27664351-27664351 on Build GRCh38
6782713 c.703G>A p.D235N Chr.2: 27664455-27664455 on Build GRCh38
7187715 c.576G>A p.G192G Chr.2: 27664328-27664328 on Build GRCh38
4093662 c.616G>A p.V206M Chr.2: 27664368-27664368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760219970 Chr.2: 27664407-27664407 on Build GRCh38
rs751999392 Chr.2: 27664424-27664424 on Build GRCh38
rs989662241 Chr.2: 27664471-27664471 on Build GRCh38
rs772860956 Chr.2: 27664371-27664371 on Build GRCh38
rs767129231 Chr.2: 27664403-27664403 on Build GRCh38
rs749484435 Chr.2: 27664443-27664443 on Build GRCh38
rs546339153 Chr.2: 27664397-27664397 on Build GRCh38
rs766748258 Chr.2: 27664419-27664419 on Build GRCh38
rs770693592 Chr.2: 27664445-27664445 on Build GRCh38
rs367822023 Chr.2: 27664425-27664425 on Build GRCh38
rs781237700 Chr.2: 27664433-27664433 on Build GRCh38
rs765503277 Chr.2: 27664298-27664298 on Build GRCh38
rs774352751 Chr.2: 27664448-27664448 on Build GRCh38
rs111609268 Chr.2: 27664393-27664393 on Build GRCh38
rs201608519 Chr.2: 27664417-27664417 on Build GRCh38
rs760059043 Chr.2: 27664351-27664351 on Build GRCh38
rs992065770 Chr.2: 27664401-27664401 on Build GRCh38
rs529018943 Chr.2: 27664320-27664320 on Build GRCh38
rs759023959 Chr.2: 27664313-27664314 on Build GRCh38
rs770652633 Chr.2: 27664396-27664396 on Build GRCh38
rs753460409 Chr.2: 27664411-27664411 on Build GRCh38
rs778017830 Chr.2: 27664439-27664439 on Build GRCh38
rs780503238 Chr.2: 27664334-27664334 on Build GRCh38
rs750844176 Chr.2: 27664310-27664310 on Build GRCh38
rs375295816 Chr.2: 27664294-27664294 on Build GRCh38
rs775194704 Chr.2: 27664406-27664406 on Build GRCh38
rs756259698 Chr.2: 27664438-27664438 on Build GRCh38
rs765677959 Chr.2: 27664391-27664391 on Build GRCh38
rs150816677 Chr.2: 27664436-27664436 on Build GRCh38
rs957118380 Chr.2: 27664382-27664382 on Build GRCh38
rs770113505 Chr.2: 27664368-27664368 on Build GRCh38
rs759548500 Chr.2: 27664400-27664400 on Build GRCh38
rs148365573 Chr.2: 27664362-27664362 on Build GRCh38
rs528154393 Chr.2: 27664276-27664276 on Build GRCh38
rs9679004 Chr.2: 27664293-27664293 on Build GRCh38
rs780696353 Chr.2: 27664423-27664424 on Build GRCh38
rs747157484 Chr.2: 27664427-27664428 on Build GRCh38
rs747540922 Chr.2: 27664350-27664350 on Build GRCh38
rs781256364 Chr.2: 27664358-27664358 on Build GRCh38
rs758797089 Chr.2: 27664315-27664315 on Build GRCh38
rs773126852 Chr.2: 27664265-27664265 on Build GRCh38
rs759067076 Chr.2: 27664291-27664291 on Build GRCh38
rs762788158 Chr.2: 27664275-27664275 on Build GRCh38

View Full Product Details