Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169895092-169895361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBR3
Entrez Gene ID
130507
Gene Name
ubiquitin protein ligase E3 component n-recognin 3 (putative)
Gene Aliases
ZNF650
UniGene
Hs.379548
Gene Chromosome Location
Chr.2: 169827463-170084131 on Build GRCh38
Transcripts
NM_172070.3 XM_005246299.4 XM_005246300.4 XM_006712268.3 XM_006712269.3 XM_006712270.3 XM_006712271.3 XM_011510604.2 XM_017003356.1 XM_017003357.1 XM_017003358.1 XM_017003359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6884762 c.1168A>G p.I390V Chr.2: 169895243-169895243 on Build GRCh38
1009379 c.1150G>T p.E384* Chr.2: 169895225-169895225 on Build GRCh38
1482180 c.1213A>G p.M405V Chr.2: 169895288-169895288 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922752535 Chr.2: 169895175-169895175 on Build GRCh38
rs566514148 Chr.2: 169895213-169895213 on Build GRCh38
rs776822388 Chr.2: 169895318-169895318 on Build GRCh38
rs376557766 Chr.2: 169895329-169895329 on Build GRCh38
rs149473540 Chr.2: 169895293-169895293 on Build GRCh38
rs763352667 Chr.2: 169895243-169895243 on Build GRCh38
rs1049805308 Chr.2: 169895131-169895131 on Build GRCh38
rs553003677 Chr.2: 169895149-169895149 on Build GRCh38
rs535452459 Chr.2: 169895230-169895230 on Build GRCh38
rs765827978 Chr.2: 169895167-169895167 on Build GRCh38
rs989203506 Chr.2: 169895170-169895170 on Build GRCh38
rs934039937 Chr.2: 169895221-169895221 on Build GRCh38
rs555257048 Chr.2: 169895281-169895281 on Build GRCh38
rs78343405 Chr.2: 169895137-169895137 on Build GRCh38
rs1049894423 Chr.2: 169895267-169895267 on Build GRCh38

View Full Product Details