Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32742614-32742853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD12
Entrez Gene ID
91646
Gene Name
tudor domain containing 12
Gene Aliases
ECAT8
UniGene
Hs.646351
Gene Chromosome Location
Chr.19: 32719773-32829580 on Build GRCh38
Transcripts
NM_001110822.1 XM_011527471.2 XM_011527472.2 XM_011527473.2 XM_011527474.2 XM_011527475.2 XM_011527476.2 XM_017027458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140489 c.326G>A p.R109Q Chr.19: 32742786-32742786 on Build GRCh38
4912345 c.383A>G p.Y128C Chr.19: 32742843-32742843 on Build GRCh38
4489032 c.341C>T p.S114L Chr.19: 32742801-32742801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950196570 Chr.19: 32742723-32742723 on Build GRCh38
rs865942933 Chr.19: 32742801-32742801 on Build GRCh38
rs191078032 Chr.19: 32742791-32742791 on Build GRCh38
rs538328148 Chr.19: 32742754-32742754 on Build GRCh38
rs898193053 Chr.19: 32742734-32742734 on Build GRCh38
rs772982873 Chr.19: 32742816-32742816 on Build GRCh38
rs1028265780 Chr.19: 32742756-32742756 on Build GRCh38
rs1045326598 Chr.19: 32742653-32742653 on Build GRCh38
rs778739637 Chr.19: 32742740-32742740 on Build GRCh38
rs905440346 Chr.19: 32742658-32742658 on Build GRCh38
rs538625433 Chr.19: 32742711-32742711 on Build GRCh38
rs1017092016 Chr.19: 32742724-32742724 on Build GRCh38
rs896082676 Chr.19: 32742714-32742714 on Build GRCh38
rs956029772 Chr.19: 32742819-32742819 on Build GRCh38
rs996902820 Chr.19: 32742750-32742750 on Build GRCh38
rs113496313 Chr.19: 32742697-32742697 on Build GRCh38
rs1047930471 Chr.19: 32742713-32742713 on Build GRCh38
rs891751891 Chr.19: 32742802-32742802 on Build GRCh38
rs748083093 Chr.19: 32742772-32742772 on Build GRCh38
rs116197390 Chr.19: 32742820-32742820 on Build GRCh38
rs758871426 Chr.19: 32742801-32742801 on Build GRCh38
rs955291592 Chr.19: 32742769-32742769 on Build GRCh38
rs939546664 Chr.19: 32742659-32742659 on Build GRCh38
rs756668790 Chr.19: 32742812-32742812 on Build GRCh38
rs764775928 Chr.19: 32742634-32742634 on Build GRCh38
rs765909374 Chr.19: 32742679-32742679 on Build GRCh38
rs918243372 Chr.19: 32742656-32742656 on Build GRCh38
rs569304889 Chr.19: 32742726-32742726 on Build GRCh38
rs558933486 Chr.19: 32742712-32742712 on Build GRCh38

View Full Product Details