Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89333013-89333286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2J1
Entrez Gene ID
51465
Gene Name
ubiquitin conjugating enzyme E2 J1
Gene Aliases
CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p
UniGene
Hs.163776
Gene Chromosome Location
Chr.6: 89326625-89352917 on Build GRCh38
Transcripts
NM_016021.2 XM_011535887.1 XM_011535888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321071 c.662C>T p.A221V Chr.6: 89333102-89333102 on Build GRCh38
1754954 c.596C>T p.S199L Chr.6: 89333168-89333168 on Build GRCh38
3178138 c.665C>T p.T222M Chr.6: 89333099-89333099 on Build GRCh38
6107567 c.677C>A p.S226* Chr.6: 89333087-89333087 on Build GRCh38
6815090 c.616delT p.S206fs*2 Chr.6: 89333148-89333148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775989219 Chr.6: 89333156-89333156 on Build GRCh38
rs748748689 Chr.6: 89333139-89333139 on Build GRCh38
rs774862010 Chr.6: 89333062-89333062 on Build GRCh38
rs754802670 Chr.6: 89333116-89333116 on Build GRCh38
rs747336938 Chr.6: 89333157-89333157 on Build GRCh38
rs200424776 Chr.6: 89333080-89333080 on Build GRCh38
rs761994024 Chr.6: 89333099-89333099 on Build GRCh38
rs763764093 Chr.6: 89333251-89333251 on Build GRCh38
rs776800982 Chr.6: 89333087-89333087 on Build GRCh38
rs746422968 Chr.6: 89333152-89333152 on Build GRCh38
rs112892732 Chr.6: 89333120-89333120 on Build GRCh38
rs780682156 Chr.6: 89333237-89333246 on Build GRCh38
rs938339185 Chr.6: 89333086-89333086 on Build GRCh38
rs754855702 Chr.6: 89333038-89333038 on Build GRCh38
rs948612481 Chr.6: 89333055-89333055 on Build GRCh38
rs777409528 Chr.6: 89333049-89333049 on Build GRCh38
rs755886423 Chr.6: 89333243-89333243 on Build GRCh38
rs771369529 Chr.6: 89333051-89333051 on Build GRCh38
rs747562729 Chr.6: 89333043-89333043 on Build GRCh38
rs761014012 Chr.6: 89333090-89333090 on Build GRCh38
rs768861274 Chr.6: 89333164-89333164 on Build GRCh38
rs767405762 Chr.6: 89333121-89333121 on Build GRCh38
rs748832646 Chr.6: 89333177-89333177 on Build GRCh38
rs376561662 Chr.6: 89333196-89333196 on Build GRCh38
rs761957092 Chr.6: 89333183-89333183 on Build GRCh38
rs752291099 Chr.6: 89333123-89333123 on Build GRCh38
rs183870615 Chr.6: 89333213-89333213 on Build GRCh38
rs994151310 Chr.6: 89333238-89333238 on Build GRCh38
rs778562635 Chr.6: 89333222-89333222 on Build GRCh38
rs764072957 Chr.6: 89333097-89333097 on Build GRCh38
rs965776244 Chr.6: 89333216-89333216 on Build GRCh38
rs371742400 Chr.6: 89333148-89333148 on Build GRCh38
rs756792961 Chr.6: 89333143-89333143 on Build GRCh38
rs745435716 Chr.6: 89333250-89333251 on Build GRCh38
rs755726697 Chr.6: 89333126-89333126 on Build GRCh38
rs144602788 Chr.6: 89333178-89333178 on Build GRCh38
rs777305881 Chr.6: 89333129-89333129 on Build GRCh38
rs374319811 Chr.6: 89333186-89333186 on Build GRCh38
rs931569260 Chr.6: 89333077-89333077 on Build GRCh38
rs769176764 Chr.6: 89333048-89333048 on Build GRCh38
rs151000241 Chr.6: 89333195-89333195 on Build GRCh38
rs759852636 Chr.6: 89333067-89333067 on Build GRCh38
rs375378658 Chr.6: 89333098-89333098 on Build GRCh38
rs201739979 Chr.6: 89333050-89333050 on Build GRCh38
rs780682197 Chr.6: 89333041-89333041 on Build GRCh38
rs772309903 Chr.6: 89333153-89333153 on Build GRCh38
rs1026176086 Chr.6: 89333258-89333258 on Build GRCh38
rs765459662 Chr.6: 89333184-89333184 on Build GRCh38
rs567236275 Chr.6: 89333115-89333115 on Build GRCh38

View Full Product Details