Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133379786-133380051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927432
Entrez Gene ID
101927432
Gene Name
uncharacterized LOC101927432
Gene Aliases
-
UniGene
Hs.665033
Gene Chromosome Location
Chr.3: 133333571-133386339 on Build GRCh38
Transcripts
NM_001136469.2 NM_001282865.1 NM_023943.3 XM_011513097.2 XM_011513098.2 XM_017007080.1 XM_017007081.1 XM_017007082.1

Gene Symbol
TMEM108
Entrez Gene ID
66000
Gene Name
transmembrane protein 108
Gene Aliases
CT124
UniGene
Hs.191616
Gene Chromosome Location
Chr.3: 133038288-133397775 on Build GRCh38
Transcripts
NM_001136469.2 NM_001282865.1 NM_023943.3 XM_011513097.2 XM_011513098.2 XM_017007080.1 XM_017007081.1 XM_017007082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7394059 c.106G>C p.E36Q Chr.3: 133379817-133379817 on Build GRCh38
728372 c.89A>G p.Q30R Chr.3: 133379800-133379800 on Build GRCh38
3122161 c.137C>T p.P46L Chr.3: 133379848-133379848 on Build GRCh38
728371 c.90G>T p.Q30H Chr.3: 133379801-133379801 on Build GRCh38
728370 c.138C>T p.P46P Chr.3: 133379849-133379849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373857528 Chr.3: 133379993-133379993 on Build GRCh38
rs533466152 Chr.3: 133380017-133380017 on Build GRCh38
rs142844774 Chr.3: 133380026-133380026 on Build GRCh38
rs755488797 Chr.3: 133379855-133379855 on Build GRCh38
rs748674531 Chr.3: 133379949-133379949 on Build GRCh38
rs758631335 Chr.3: 133379929-133379929 on Build GRCh38
rs780460288 Chr.3: 133379847-133379847 on Build GRCh38
rs149345637 Chr.3: 133379817-133379817 on Build GRCh38
rs779983086 Chr.3: 133379898-133379898 on Build GRCh38
rs368821010 Chr.3: 133379892-133379892 on Build GRCh38
rs567986332 Chr.3: 133380016-133380016 on Build GRCh38
rs372281436 Chr.3: 133379922-133379922 on Build GRCh38
rs760440546 Chr.3: 133379914-133379914 on Build GRCh38
rs1028916806 Chr.3: 133379814-133379814 on Build GRCh38
rs966321812 Chr.3: 133379854-133379854 on Build GRCh38
rs893019417 Chr.3: 133379975-133379975 on Build GRCh38
rs755326269 Chr.3: 133379947-133379947 on Build GRCh38
rs143911768 Chr.3: 133380027-133380027 on Build GRCh38
rs756757516 Chr.3: 133379955-133379955 on Build GRCh38
rs34111099 Chr.3: 133379962-133379962 on Build GRCh38
rs1022407053 Chr.3: 133379832-133379832 on Build GRCh38
rs770584065 Chr.3: 133379873-133379873 on Build GRCh38
rs200624038 Chr.3: 133379845-133379845 on Build GRCh38
rs746780009 Chr.3: 133379984-133379984 on Build GRCh38
rs1055718719 Chr.3: 133379963-133379963 on Build GRCh38
rs1018937358 Chr.3: 133379852-133379852 on Build GRCh38
rs773319249 Chr.3: 133380012-133380012 on Build GRCh38
rs745541442 Chr.3: 133379891-133379891 on Build GRCh38
rs774086467 Chr.3: 133379887-133379887 on Build GRCh38
rs751920405 Chr.3: 133379945-133379945 on Build GRCh38
rs376694463 Chr.3: 133379864-133379864 on Build GRCh38
rs769779341 Chr.3: 133380011-133380011 on Build GRCh38
rs202042493 Chr.3: 133379928-133379928 on Build GRCh38
rs966756695 Chr.3: 133379837-133379837 on Build GRCh38
rs952247572 Chr.3: 133379911-133379911 on Build GRCh38
rs564663344 Chr.3: 133379869-133379869 on Build GRCh38
rs776494932 Chr.3: 133379997-133379997 on Build GRCh38
rs771692595 Chr.3: 133379893-133379893 on Build GRCh38
rs761732961 Chr.3: 133380008-133380008 on Build GRCh38
rs1044355156 Chr.3: 133379969-133379969 on Build GRCh38
rs767996292 Chr.3: 133380019-133380019 on Build GRCh38
rs745430893 Chr.3: 133379971-133379971 on Build GRCh38
rs148532842 Chr.3: 133379938-133379938 on Build GRCh38
rs548135838 Chr.3: 133379978-133379978 on Build GRCh38
rs199598943 Chr.3: 133379921-133379921 on Build GRCh38
rs781744470 Chr.3: 133379948-133379948 on Build GRCh38
rs928298661 Chr.3: 133379956-133379956 on Build GRCh38
rs138079048 Chr.3: 133380014-133380014 on Build GRCh38
rs763987660 Chr.3: 133379919-133379919 on Build GRCh38
rs147436918 Chr.3: 133379981-133379981 on Build GRCh38
rs930518082 Chr.3: 133379970-133379970 on Build GRCh38
rs747390187 Chr.3: 133379851-133379851 on Build GRCh38
rs778125233 Chr.3: 133380027-133380028 on Build GRCh38

View Full Product Details