Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47915744-47915971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF81
Entrez Gene ID
347344
Gene Name
zinc finger protein 81
Gene Aliases
HFZ20, MRX45, dJ54B20.6
UniGene
Hs.114246
Gene Chromosome Location
Chr.X: 47836864-47925626 on Build GRCh38
Transcripts
NM_007137.3 XM_005272600.3 XM_011543899.2 XM_011543900.2 XM_017029486.1 XM_017029487.1

SNP Detail

SNP ID Chromosome Location
rs782333599 Chr.X: 47915900-47915900 on Build GRCh38
rs371224084 Chr.X: 47915796-47915796 on Build GRCh38
rs782377735 Chr.X: 47915829-47915829 on Build GRCh38
rs782138015 Chr.X: 47915929-47915929 on Build GRCh38
rs782218495 Chr.X: 47915879-47915879 on Build GRCh38
rs782202189 Chr.X: 47915918-47915918 on Build GRCh38
rs200890712 Chr.X: 47915910-47915910 on Build GRCh38
rs988101883 Chr.X: 47915896-47915896 on Build GRCh38
rs912442591 Chr.X: 47915937-47915937 on Build GRCh38
rs781972062 Chr.X: 47915862-47915862 on Build GRCh38
rs782566228 Chr.X: 47915803-47915803 on Build GRCh38

View Full Product Details