Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52707037-52707288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC6
Entrez Gene ID
5706
Gene Name
proteasome 26S subunit, ATPase 6
Gene Aliases
CADP44, HEL-S-73, P44, SUG2, p42
UniGene
Hs.156171
Gene Chromosome Location
Chr.14: 52707163-52727998 on Build GRCh38
Transcripts
NM_002806.3 XM_017021472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7111597 c.42C>A p.I14I Chr.14: 52707219-52707219 on Build GRCh38
3496391 c.18C>T p.I6I Chr.14: 52707195-52707195 on Build GRCh38
433130 c.2T>C p.M1T Chr.14: 52707179-52707179 on Build GRCh38
41490 c.63G>T p.E21D Chr.14: 52707282-52707282 on Build GRCh38
384289 c.7G>C p.D3H Chr.14: 52707226-52707226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371829618 Chr.14: 52707211-52707211 on Build GRCh38
rs142367793 Chr.14: 52707229-52707229 on Build GRCh38
rs899448406 Chr.14: 52707192-52707192 on Build GRCh38
rs972391597 Chr.14: 52707061-52707061 on Build GRCh38
rs764840411 Chr.14: 52707134-52707134 on Build GRCh38
rs554296759 Chr.14: 52707074-52707074 on Build GRCh38
rs539034817 Chr.14: 52707112-52707112 on Build GRCh38
rs140545868 Chr.14: 52707202-52707202 on Build GRCh38
rs780003575 Chr.14: 52707093-52707093 on Build GRCh38
rs765176425 Chr.14: 52707228-52707228 on Build GRCh38
rs750227004 Chr.14: 52707181-52707181 on Build GRCh38
rs534271494 Chr.14: 52707071-52707071 on Build GRCh38
rs151244912 Chr.14: 52707256-52707256 on Build GRCh38
rs577375404 Chr.14: 52707110-52707110 on Build GRCh38
rs777973212 Chr.14: 52707236-52707236 on Build GRCh38
rs781501702 Chr.14: 52707087-52707087 on Build GRCh38
rs750143693 Chr.14: 52707138-52707138 on Build GRCh38
rs371262723 Chr.14: 52707225-52707225 on Build GRCh38
rs769953707 Chr.14: 52707194-52707194 on Build GRCh38
rs970560977 Chr.14: 52707064-52707064 on Build GRCh38
rs911390148 Chr.14: 52707073-52707073 on Build GRCh38
rs757175270 Chr.14: 52707174-52707174 on Build GRCh38
rs1034646372 Chr.14: 52707118-52707118 on Build GRCh38
rs771076814 Chr.14: 52707242-52707242 on Build GRCh38
rs867983128 Chr.14: 52707164-52707164 on Build GRCh38
rs376129677 Chr.14: 52707214-52707214 on Build GRCh38
rs758135778 Chr.14: 52707182-52707182 on Build GRCh38
rs199988613 Chr.14: 52707216-52707216 on Build GRCh38
rs201442969 Chr.14: 52707146-52707146 on Build GRCh38
rs769717259 Chr.14: 52707133-52707133 on Build GRCh38
rs779585393 Chr.14: 52707248-52707248 on Build GRCh38
rs748428557 Chr.14: 52707258-52707259 on Build GRCh38
rs542227590 Chr.14: 52707235-52707235 on Build GRCh38
rs140470588 Chr.14: 52707264-52707264 on Build GRCh38
rs753343535 Chr.14: 52707213-52707213 on Build GRCh38
rs753009195 Chr.14: 52707233-52707233 on Build GRCh38
rs537400605 Chr.14: 52707202-52707203 on Build GRCh38
rs879132794 Chr.14: 52707122-52707122 on Build GRCh38
rs778748819 Chr.14: 52707175-52707175 on Build GRCh38
rs773795719 Chr.14: 52707141-52707141 on Build GRCh38
rs779678774 Chr.14: 52707231-52707231 on Build GRCh38
rs752066600 Chr.14: 52707161-52707161 on Build GRCh38
rs749384722 Chr.14: 52707238-52707238 on Build GRCh38
rs999793856 Chr.14: 52707247-52707247 on Build GRCh38
rs751705678 Chr.14: 52707059-52707059 on Build GRCh38
rs765820567 Chr.14: 52707142-52707143 on Build GRCh38
rs781503856 Chr.14: 52707190-52707190 on Build GRCh38
rs984811268 Chr.14: 52707072-52707072 on Build GRCh38
rs777689414 Chr.14: 52707195-52707195 on Build GRCh38
rs767203062 Chr.14: 52707162-52707162 on Build GRCh38
rs771558844 Chr.14: 52707198-52707198 on Build GRCh38
rs894945610 Chr.14: 52707178-52707178 on Build GRCh38
rs772535111 Chr.14: 52707260-52707260 on Build GRCh38
rs921698451 Chr.14: 52707086-52707086 on Build GRCh38
rs759992296 Chr.14: 52707203-52707203 on Build GRCh38
rs753864495 Chr.14: 52707173-52707173 on Build GRCh38
rs775248842 Chr.14: 52707208-52707208 on Build GRCh38
rs774841132 Chr.14: 52707148-52707148 on Build GRCh38
rs371360008 Chr.14: 52707170-52707170 on Build GRCh38
rs755461560 Chr.14: 52707160-52707161 on Build GRCh38
rs190752409 Chr.14: 52707065-52707065 on Build GRCh38
rs747915837 Chr.14: 52707166-52707166 on Build GRCh38
rs749865994 Chr.14: 52707197-52707197 on Build GRCh38
rs1041720729 Chr.14: 52707106-52707106 on Build GRCh38
rs894485259 Chr.14: 52707200-52707200 on Build GRCh38
rs923876828 Chr.14: 52707066-52707066 on Build GRCh38
rs539907616 Chr.14: 52707142-52707142 on Build GRCh38
rs942863250 Chr.14: 52707084-52707085 on Build GRCh38
rs757892543 Chr.14: 52707139-52707140 on Build GRCh38
rs946006658 Chr.14: 52707089-52707089 on Build GRCh38
rs755251197 Chr.14: 52707070-52707070 on Build GRCh38
rs759327634 Chr.14: 52707152-52707152 on Build GRCh38
rs748273413 Chr.14: 52707193-52707193 on Build GRCh38
rs772275442 Chr.14: 52707207-52707207 on Build GRCh38

View Full Product Details