Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62467138-62467363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO11
Entrez Gene ID
51194
Gene Name
importin 11
Gene Aliases
RanBP11
UniGene
Hs.482269
Gene Chromosome Location
Chr.5: 62412746-62628589 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6686643 c.719A>T p.N240I Chr.5: 62467213-62467213 on Build GRCh38
5737238 c.681C>A p.H227Q Chr.5: 62467175-62467175 on Build GRCh38
3429544 c.662C>T p.A221V Chr.5: 62467156-62467156 on Build GRCh38
3828263 c.713C>G p.S238C Chr.5: 62467207-62467207 on Build GRCh38
3828264 c.593C>G p.S198C Chr.5: 62467207-62467207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755783411 Chr.5: 62467297-62467297 on Build GRCh38
rs952035141 Chr.5: 62467191-62467191 on Build GRCh38
rs776903274 Chr.5: 62467267-62467267 on Build GRCh38
rs772998136 Chr.5: 62467246-62467246 on Build GRCh38
rs764373877 Chr.5: 62467287-62467287 on Build GRCh38
rs377471750 Chr.5: 62467279-62467279 on Build GRCh38
rs748745011 Chr.5: 62467230-62467230 on Build GRCh38
rs753383959 Chr.5: 62467304-62467304 on Build GRCh38
rs763463619 Chr.5: 62467283-62467283 on Build GRCh38
rs980179147 Chr.5: 62467274-62467274 on Build GRCh38
rs927384977 Chr.5: 62467286-62467286 on Build GRCh38
rs139195186 Chr.5: 62467253-62467253 on Build GRCh38
rs753131279 Chr.5: 62467282-62467282 on Build GRCh38
rs370052124 Chr.5: 62467213-62467213 on Build GRCh38
rs771235982 Chr.5: 62467256-62467256 on Build GRCh38
rs780713008 Chr.5: 62467267-62467275 on Build GRCh38
rs540919449 Chr.5: 62467336-62467336 on Build GRCh38
rs866866734 Chr.5: 62467248-62467248 on Build GRCh38
rs778683858 Chr.5: 62467242-62467242 on Build GRCh38
rs779572606 Chr.5: 62467218-62467218 on Build GRCh38
rs759660821 Chr.5: 62467269-62467269 on Build GRCh38
rs373208617 Chr.5: 62467187-62467187 on Build GRCh38
rs376400435 Chr.5: 62467210-62467210 on Build GRCh38
rs780956596 Chr.5: 62467205-62467205 on Build GRCh38
rs374430847 Chr.5: 62467238-62467238 on Build GRCh38
rs949869381 Chr.5: 62467281-62467281 on Build GRCh38
rs771746187 Chr.5: 62467243-62467243 on Build GRCh38
rs754476276 Chr.5: 62467312-62467312 on Build GRCh38
rs778966632 Chr.5: 62467313-62467313 on Build GRCh38
rs112836279 Chr.5: 62467322-62467322 on Build GRCh38
rs1042331532 Chr.5: 62467337-62467337 on Build GRCh38
rs751914306 Chr.5: 62467291-62467291 on Build GRCh38
rs766576355 Chr.5: 62467179-62467179 on Build GRCh38
rs779656552 Chr.5: 62467302-62467302 on Build GRCh38
rs368808112 Chr.5: 62467301-62467301 on Build GRCh38
rs753149976 Chr.5: 62467190-62467190 on Build GRCh38
rs760145602 Chr.5: 62467186-62467186 on Build GRCh38
rs747836354 Chr.5: 62467288-62467289 on Build GRCh38
rs367843719 Chr.5: 62467325-62467325 on Build GRCh38
rs753900312 Chr.5: 62467184-62467184 on Build GRCh38
rs758689101 Chr.5: 62467196-62467196 on Build GRCh38
rs571439807 Chr.5: 62467236-62467236 on Build GRCh38

View Full Product Details