Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67181241-67181466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_011544860.2 XM_017017391.1 XM_017017392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451233 c.187A>G p.N63D Chr.11: 67181325-67181325 on Build GRCh38
3452247 c.215T>A p.L72* Chr.11: 67181353-67181353 on Build GRCh38
5899033 c.242C>T p.S81F Chr.11: 67181380-67181380 on Build GRCh38
7373321 c.244G>C p.D82H Chr.11: 67181382-67181382 on Build GRCh38
5891440 c.190G>T p.V64L Chr.11: 67181328-67181328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763216277 Chr.11: 67181445-67181446 on Build GRCh38
rs78354165 Chr.11: 67181433-67181433 on Build GRCh38
rs767525906 Chr.11: 67181413-67181413 on Build GRCh38
rs754577818 Chr.11: 67181287-67181287 on Build GRCh38
rs753518729 Chr.11: 67181285-67181285 on Build GRCh38
rs773239884 Chr.11: 67181417-67181417 on Build GRCh38
rs906973652 Chr.11: 67181424-67181424 on Build GRCh38
rs370667174 Chr.11: 67181442-67181442 on Build GRCh38
rs371032458 Chr.11: 67181307-67181307 on Build GRCh38
rs886909974 Chr.11: 67181418-67181418 on Build GRCh38
rs775807056 Chr.11: 67181292-67181295 on Build GRCh38
rs775656106 Chr.11: 67181377-67181377 on Build GRCh38
rs746056503 Chr.11: 67181347-67181347 on Build GRCh38
rs762876792 Chr.11: 67181390-67181390 on Build GRCh38
rs755936661 Chr.11: 67181282-67181282 on Build GRCh38
rs771232627 Chr.11: 67181321-67181321 on Build GRCh38
rs919877211 Chr.11: 67181310-67181310 on Build GRCh38
rs559111143 Chr.11: 67181431-67181431 on Build GRCh38
rs377444009 Chr.11: 67181313-67181313 on Build GRCh38
rs373536076 Chr.11: 67181360-67181360 on Build GRCh38
rs371496319 Chr.11: 67181342-67181342 on Build GRCh38
rs776883963 Chr.11: 67181328-67181328 on Build GRCh38
rs374354689 Chr.11: 67181443-67181443 on Build GRCh38
rs778483788 Chr.11: 67181293-67181293 on Build GRCh38
rs778074773 Chr.11: 67181312-67181312 on Build GRCh38
rs766152981 Chr.11: 67181283-67181283 on Build GRCh38
rs750252227 Chr.11: 67181272-67181272 on Build GRCh38
rs748659600 Chr.11: 67181298-67181298 on Build GRCh38
rs1046916274 Chr.11: 67181345-67181345 on Build GRCh38
rs986646054 Chr.11: 67181315-67181315 on Build GRCh38

View Full Product Details