Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38688023-38688276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM2
Entrez Gene ID
341640
Gene Name
FRAS1 related extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.253994
Gene Chromosome Location
Chr.13: 38687036-38887131 on Build GRCh38
Transcripts
NM_207361.5 XM_017020554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622822 c.839T>C p.I280T Chr.13: 38688183-38688183 on Build GRCh38
7153680 c.923G>T p.R308M Chr.13: 38688267-38688267 on Build GRCh38
6666864 c.747G>A p.P249P Chr.13: 38688091-38688091 on Build GRCh38
5498104 c.796T>C p.Y266H Chr.13: 38688140-38688140 on Build GRCh38
6666865 c.742G>A p.A248T Chr.13: 38688086-38688086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748783790 Chr.13: 38688155-38688155 on Build GRCh38
rs547302858 Chr.13: 38688080-38688080 on Build GRCh38
rs539273741 Chr.13: 38688151-38688151 on Build GRCh38
rs763205800 Chr.13: 38688117-38688117 on Build GRCh38
rs952813625 Chr.13: 38688158-38688158 on Build GRCh38
rs766868798 Chr.13: 38688243-38688243 on Build GRCh38
rs982499550 Chr.13: 38688215-38688215 on Build GRCh38
rs752025828 Chr.13: 38688131-38688131 on Build GRCh38
rs756672907 Chr.13: 38688236-38688236 on Build GRCh38
rs750877781 Chr.13: 38688184-38688185 on Build GRCh38
rs746871746 Chr.13: 38688179-38688179 on Build GRCh38
rs779482351 Chr.13: 38688154-38688154 on Build GRCh38
rs1019883498 Chr.13: 38688090-38688090 on Build GRCh38
rs748873581 Chr.13: 38688055-38688055 on Build GRCh38
rs755354281 Chr.13: 38688054-38688054 on Build GRCh38
rs757542294 Chr.13: 38688133-38688133 on Build GRCh38
rs778649414 Chr.13: 38688257-38688257 on Build GRCh38
rs976958580 Chr.13: 38688118-38688118 on Build GRCh38
rs200898675 Chr.13: 38688170-38688170 on Build GRCh38
rs774390073 Chr.13: 38688210-38688210 on Build GRCh38
rs1016062641 Chr.13: 38688188-38688188 on Build GRCh38
rs779414030 Chr.13: 38688173-38688174 on Build GRCh38
rs781523250 Chr.13: 38688043-38688043 on Build GRCh38
rs570395955 Chr.13: 38688105-38688105 on Build GRCh38
rs750941575 Chr.13: 38688223-38688223 on Build GRCh38
rs752967423 Chr.13: 38688114-38688114 on Build GRCh38
rs901396989 Chr.13: 38688121-38688121 on Build GRCh38
rs745435644 Chr.13: 38688044-38688044 on Build GRCh38
rs753388275 Chr.13: 38688253-38688253 on Build GRCh38
rs796251550 Chr.13: 38688057-38688058 on Build GRCh38
rs781735102 Chr.13: 38688134-38688134 on Build GRCh38
rs1032260079 Chr.13: 38688046-38688046 on Build GRCh38
rs772999478 Chr.13: 38688068-38688068 on Build GRCh38
rs374073091 Chr.13: 38688221-38688221 on Build GRCh38
rs555193496 Chr.13: 38688059-38688059 on Build GRCh38
rs199563252 Chr.13: 38688256-38688256 on Build GRCh38
rs757008811 Chr.13: 38688114-38688122 on Build GRCh38
rs762272605 Chr.13: 38688218-38688218 on Build GRCh38
rs774262015 Chr.13: 38688061-38688061 on Build GRCh38
rs761511094 Chr.13: 38688064-38688064 on Build GRCh38
rs769249087 Chr.13: 38688049-38688049 on Build GRCh38
rs116236946 Chr.13: 38688058-38688058 on Build GRCh38
rs998887205 Chr.13: 38688071-38688071 on Build GRCh38
rs764395172 Chr.13: 38688120-38688120 on Build GRCh38
rs369046419 Chr.13: 38688150-38688150 on Build GRCh38
rs201675703 Chr.13: 38688162-38688162 on Build GRCh38
rs770004356 Chr.13: 38688194-38688194 on Build GRCh38
rs370369074 Chr.13: 38688177-38688177 on Build GRCh38
rs772085604 Chr.13: 38688067-38688067 on Build GRCh38
rs748022903 Chr.13: 38688169-38688169 on Build GRCh38
rs997045696 Chr.13: 38688127-38688127 on Build GRCh38
rs764428476 Chr.13: 38688207-38688207 on Build GRCh38
rs775767577 Chr.13: 38688202-38688202 on Build GRCh38
rs763117886 Chr.13: 38688203-38688203 on Build GRCh38

View Full Product Details