Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28237806-28238031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
UniGene
Hs.744914
Gene Chromosome Location
Chr.1: 28236091-28238105 on Build GRCh38
Transcripts
NM_016311.4 NM_178190.2 NM_178191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907962 c.297C>T p.I99I Chr.1: 28237954-28237954 on Build GRCh38
7223833 c.280C>T p.R94C Chr.1: 28237937-28237937 on Build GRCh38
6608459 c.235G>A p.V79I Chr.1: 28237892-28237892 on Build GRCh38
5380574 c.286A>G p.K96E Chr.1: 28237943-28237943 on Build GRCh38
5380573 c.262C>T p.L88L Chr.1: 28237919-28237919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776903465 Chr.1: 28237963-28237963 on Build GRCh38
rs139876260 Chr.1: 28237955-28237955 on Build GRCh38
rs755576959 Chr.1: 28237910-28237910 on Build GRCh38
rs746429737 Chr.1: 28237901-28237903 on Build GRCh38
rs374584642 Chr.1: 28237937-28237937 on Build GRCh38
rs777305106 Chr.1: 28237911-28237911 on Build GRCh38
rs754730947 Chr.1: 28237901-28237901 on Build GRCh38
rs112006347 Chr.1: 28237835-28237835 on Build GRCh38
rs767244362 Chr.1: 28237997-28237997 on Build GRCh38
rs747774169 Chr.1: 28237906-28237906 on Build GRCh38
rs762972864 Chr.1: 28237968-28237968 on Build GRCh38
rs780836240 Chr.1: 28237903-28237903 on Build GRCh38
rs771483926 Chr.1: 28237916-28237916 on Build GRCh38
rs772338600 Chr.1: 28237935-28237935 on Build GRCh38
rs752359232 Chr.1: 28238009-28238009 on Build GRCh38
rs760751368 Chr.1: 28237865-28237865 on Build GRCh38
rs1045059725 Chr.1: 28237946-28237947 on Build GRCh38
rs959763996 Chr.1: 28237921-28237921 on Build GRCh38
rs113261974 Chr.1: 28237834-28237834 on Build GRCh38
rs754750123 Chr.1: 28237988-28237988 on Build GRCh38
rs879748592 Chr.1: 28237876-28237876 on Build GRCh38
rs201956032 Chr.1: 28237897-28237897 on Build GRCh38
rs756993246 Chr.1: 28238007-28238008 on Build GRCh38
rs758401235 Chr.1: 28237892-28237892 on Build GRCh38
rs746179050 Chr.1: 28237926-28237926 on Build GRCh38
rs755666933 Chr.1: 28237923-28237926 on Build GRCh38
rs763390750 Chr.1: 28237833-28237844 on Build GRCh38
rs763719665 Chr.1: 28237947-28237949 on Build GRCh38
rs951938160 Chr.1: 28237900-28237900 on Build GRCh38
rs764292791 Chr.1: 28237884-28237884 on Build GRCh38
rs995815918 Chr.1: 28237891-28237891 on Build GRCh38
rs570378650 Chr.1: 28237845-28237845 on Build GRCh38
rs1055776 Chr.1: 28237985-28237985 on Build GRCh38
rs761008954 Chr.1: 28237938-28237938 on Build GRCh38
rs1027287407 Chr.1: 28237898-28237898 on Build GRCh38
rs556914230 Chr.1: 28237917-28237917 on Build GRCh38
rs752869401 Chr.1: 28237846-28237846 on Build GRCh38
rs751431301 Chr.1: 28237899-28237899 on Build GRCh38
rs983352469 Chr.1: 28237925-28237925 on Build GRCh38
rs199945228 Chr.1: 28237970-28237970 on Build GRCh38
rs753551798 Chr.1: 28237968-28237970 on Build GRCh38
rs749771670 Chr.1: 28237914-28237914 on Build GRCh38
rs767011179 Chr.1: 28237874-28237876 on Build GRCh38

View Full Product Details