Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34085995-34086256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN1
Entrez Gene ID
4076
Gene Name
cell cycle associated protein 1
Gene Aliases
GPIAP1, GPIP137, GRIP137, M11S1, RNG105, p137GPI
UniGene
Hs.471818
Gene Chromosome Location
Chr.11: 34051617-34102610 on Build GRCh38
Transcripts
NM_005898.4 NM_203364.2 XM_017017749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687272 c.1054C>G p.P352A Chr.11: 34086151-34086151 on Build GRCh38
250135 c.1043C>T p.A348V Chr.11: 34086140-34086140 on Build GRCh38
3687271 c.1054C>G p.P352A Chr.11: 34086151-34086151 on Build GRCh38
1703798 c.1055C>T p.P352L Chr.11: 34086152-34086152 on Build GRCh38
6421616 c.1021C>A p.P341T Chr.11: 34086118-34086118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759010901 Chr.11: 34086189-34086189 on Build GRCh38
rs746713293 Chr.11: 34086031-34086031 on Build GRCh38
rs1013625698 Chr.11: 34086062-34086062 on Build GRCh38
rs764419574 Chr.11: 34086149-34086149 on Build GRCh38
rs778432679 Chr.11: 34086019-34086019 on Build GRCh38
rs779598606 Chr.11: 34086028-34086028 on Build GRCh38
rs112338854 Chr.11: 34086059-34086059 on Build GRCh38
rs751085037 Chr.11: 34086216-34086216 on Build GRCh38
rs142679290 Chr.11: 34086103-34086103 on Build GRCh38
rs768408806 Chr.11: 34086032-34086032 on Build GRCh38
rs1008930453 Chr.11: 34086049-34086049 on Build GRCh38
rs201906028 Chr.11: 34086095-34086095 on Build GRCh38
rs772999570 Chr.11: 34086132-34086132 on Build GRCh38
rs767010881 Chr.11: 34086046-34086047 on Build GRCh38
rs776478836 Chr.11: 34086034-34086034 on Build GRCh38
rs769722711 Chr.11: 34086127-34086127 on Build GRCh38
rs779453799 Chr.11: 34086104-34086105 on Build GRCh38
rs754583192 Chr.11: 34086223-34086223 on Build GRCh38
rs750355558 Chr.11: 34086121-34086121 on Build GRCh38
rs767683678 Chr.11: 34086057-34086057 on Build GRCh38
rs377578608 Chr.11: 34086064-34086064 on Build GRCh38
rs749284381 Chr.11: 34086133-34086133 on Build GRCh38
rs774448083 Chr.11: 34086046-34086046 on Build GRCh38
rs755452485 Chr.11: 34086054-34086055 on Build GRCh38
rs762294661 Chr.11: 34086166-34086166 on Build GRCh38
rs1044673406 Chr.11: 34086079-34086081 on Build GRCh38
rs113256282 Chr.11: 34086225-34086225 on Build GRCh38
rs934027218 Chr.11: 34086017-34086017 on Build GRCh38
rs772036228 Chr.11: 34086134-34086134 on Build GRCh38
rs750864973 Chr.11: 34086185-34086185 on Build GRCh38
rs753054515 Chr.11: 34086107-34086112 on Build GRCh38
rs754685034 Chr.11: 34086115-34086115 on Build GRCh38
rs1132972 Chr.11: 34086205-34086205 on Build GRCh38
rs760918575 Chr.11: 34086144-34086144 on Build GRCh38
rs771719841 Chr.11: 34086026-34086026 on Build GRCh38
rs575272575 Chr.11: 34086154-34086154 on Build GRCh38
rs890447308 Chr.11: 34086041-34086041 on Build GRCh38
rs757774847 Chr.11: 34086067-34086067 on Build GRCh38
rs757828401 Chr.11: 34086098-34086098 on Build GRCh38
rs370502113 Chr.11: 34086114-34086114 on Build GRCh38
rs765697606 Chr.11: 34086094-34086094 on Build GRCh38
rs941519208 Chr.11: 34086042-34086042 on Build GRCh38
rs993950993 Chr.11: 34086027-34086027 on Build GRCh38
rs200204494 Chr.11: 34086055-34086055 on Build GRCh38
rs765795853 Chr.11: 34086195-34086195 on Build GRCh38
rs1030728413 Chr.11: 34086157-34086157 on Build GRCh38
rs529818858 Chr.11: 34086037-34086037 on Build GRCh38
rs745376042 Chr.11: 34086021-34086021 on Build GRCh38
rs749938795 Chr.11: 34086047-34086047 on Build GRCh38
rs775767681 Chr.11: 34086138-34086138 on Build GRCh38
rs765686887 Chr.11: 34086175-34086175 on Build GRCh38
rs200891413 Chr.11: 34086040-34086040 on Build GRCh38
rs777097874 Chr.11: 34086152-34086152 on Build GRCh38

View Full Product Details