Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110282408-110282681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A2
Entrez Gene ID
488
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Gene Aliases
ATP2B, DAR, DD, SERCA2
UniGene
Hs.506759
Gene Chromosome Location
Chr.12: 110281227-110351093 on Build GRCh38
Transcripts
NM_001681.3 NM_170665.3 XM_005253888.2 XM_011538402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467775 c.136+1G>C p.? Chr.12: 110282622-110282622 on Build GRCh38
6326063 c.119-3C>T p.? Chr.12: 110282601-110282601 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs151294187 Chr.12: 110282614-110282614 on Build GRCh38
rs114726034 Chr.12: 110282508-110282508 on Build GRCh38
rs777008704 Chr.12: 110282633-110282633 on Build GRCh38
rs758407356 Chr.12: 110282610-110282610 on Build GRCh38
rs372885221 Chr.12: 110282565-110282565 on Build GRCh38
rs540446116 Chr.12: 110282539-110282540 on Build GRCh38
rs997965600 Chr.12: 110282549-110282549 on Build GRCh38
rs763637617 Chr.12: 110282641-110282642 on Build GRCh38
rs910713443 Chr.12: 110282641-110282641 on Build GRCh38
rs993473245 Chr.12: 110282460-110282460 on Build GRCh38
rs769052817 Chr.12: 110282632-110282632 on Build GRCh38
rs780672760 Chr.12: 110282611-110282611 on Build GRCh38
rs573952845 Chr.12: 110282434-110282434 on Build GRCh38
rs757578349 Chr.12: 110282572-110282572 on Build GRCh38
rs779194575 Chr.12: 110282487-110282487 on Build GRCh38
rs978448254 Chr.12: 110282435-110282435 on Build GRCh38
rs386352281 Chr.12: 110282616-110282616 on Build GRCh38
rs183257438 Chr.12: 110282579-110282579 on Build GRCh38
rs1049041700 Chr.12: 110282539-110282542 on Build GRCh38
rs369478008 Chr.12: 110282555-110282555 on Build GRCh38
rs879476746 Chr.12: 110282547-110282547 on Build GRCh38
rs565878321 Chr.12: 110282448-110282448 on Build GRCh38
rs745965988 Chr.12: 110282593-110282593 on Build GRCh38
rs760125967 Chr.12: 110282629-110282630 on Build GRCh38
rs1004941830 Chr.12: 110282546-110282547 on Build GRCh38
rs757531836 Chr.12: 110282441-110282442 on Build GRCh38
rs748460765 Chr.12: 110282654-110282654 on Build GRCh38
rs1012756151 Chr.12: 110282490-110282490 on Build GRCh38
rs753785245 Chr.12: 110282644-110282645 on Build GRCh38
rs887659532 Chr.12: 110282462-110282462 on Build GRCh38
rs779818256 Chr.12: 110282545-110282548 on Build GRCh38
rs1027586263 Chr.12: 110282461-110282461 on Build GRCh38
rs925717503 Chr.12: 110282458-110282458 on Build GRCh38
rs755305838 Chr.12: 110282478-110282478 on Build GRCh38
rs191058402 Chr.12: 110282517-110282517 on Build GRCh38
rs767396858 Chr.12: 110282560-110282560 on Build GRCh38
rs752355242 Chr.12: 110282573-110282574 on Build GRCh38
rs1032649497 Chr.12: 110282601-110282601 on Build GRCh38
rs930679598 Chr.12: 110282525-110282525 on Build GRCh38
rs769496904 Chr.12: 110282657-110282657 on Build GRCh38
rs372941910 Chr.12: 110282557-110282558 on Build GRCh38
rs188083000 Chr.12: 110282493-110282493 on Build GRCh38
rs955726400 Chr.12: 110282524-110282524 on Build GRCh38
rs900976488 Chr.12: 110282548-110282548 on Build GRCh38
rs111631659 Chr.12: 110282522-110282522 on Build GRCh38

View Full Product Details