Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49257329-49257602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs782264212 Chr.X: 49257482-49257482 on Build GRCh38
rs782528935 Chr.X: 49257447-49257447 on Build GRCh38
rs1033545455 Chr.X: 49257509-49257509 on Build GRCh38
rs782158863 Chr.X: 49257412-49257412 on Build GRCh38
rs782691809 Chr.X: 49257573-49257573 on Build GRCh38
rs781892250 Chr.X: 49257411-49257411 on Build GRCh38
rs782124230 Chr.X: 49257549-49257549 on Build GRCh38
rs782063944 Chr.X: 49257386-49257386 on Build GRCh38
rs781831285 Chr.X: 49257548-49257548 on Build GRCh38
rs782303740 Chr.X: 49257540-49257540 on Build GRCh38
rs782665522 Chr.X: 49257478-49257478 on Build GRCh38
rs782776637 Chr.X: 49257438-49257438 on Build GRCh38
rs782820440 Chr.X: 49257358-49257358 on Build GRCh38
rs782604073 Chr.X: 49257489-49257489 on Build GRCh38
rs782511378 Chr.X: 49257483-49257483 on Build GRCh38
rs56409830 Chr.X: 49257518-49257518 on Build GRCh38
rs1002017383 Chr.X: 49257484-49257484 on Build GRCh38
rs782550900 Chr.X: 49257475-49257475 on Build GRCh38
rs782693232 Chr.X: 49257403-49257403 on Build GRCh38
rs782086170 Chr.X: 49257568-49257568 on Build GRCh38
rs143632471 Chr.X: 49257545-49257545 on Build GRCh38
rs782202711 Chr.X: 49257505-49257505 on Build GRCh38
rs781944242 Chr.X: 49257376-49257376 on Build GRCh38
rs200554980 Chr.X: 49257541-49257541 on Build GRCh38
rs781856708 Chr.X: 49257442-49257442 on Build GRCh38
rs56239837 Chr.X: 49257374-49257374 on Build GRCh38
rs977920377 Chr.X: 49257569-49257569 on Build GRCh38
rs782640594 Chr.X: 49257520-49257520 on Build GRCh38
rs782613398 Chr.X: 49257473-49257473 on Build GRCh38
rs782572328 Chr.X: 49257558-49257558 on Build GRCh38

View Full Product Details