Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54477971-54478204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFB
Entrez Gene ID
2764
Gene Name
glia maturation factor beta
Gene Aliases
GMF
UniGene
Hs.151413
Gene Chromosome Location
Chr.14: 54474491-54489026 on Build GRCh38
Transcripts
NM_004124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263193 c.403C>T p.R135C Chr.14: 54478114-54478114 on Build GRCh38
5943987 c.357-4delT p.? Chr.14: 54478164-54478164 on Build GRCh38
4051327 c.379G>A p.E127K Chr.14: 54478138-54478138 on Build GRCh38
6240385 c.357-4_357-3insT p.? Chr.14: 54478163-54478164 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774446977 Chr.14: 54478055-54478055 on Build GRCh38
rs749331288 Chr.14: 54478178-54478178 on Build GRCh38
rs756735322 Chr.14: 54478099-54478099 on Build GRCh38
rs770243322 Chr.14: 54478150-54478150 on Build GRCh38
rs748534054 Chr.14: 54478147-54478147 on Build GRCh38
rs78616058 Chr.14: 54478110-54478110 on Build GRCh38
rs781515004 Chr.14: 54478145-54478145 on Build GRCh38
rs571831252 Chr.14: 54478170-54478170 on Build GRCh38
rs371009786 Chr.14: 54478114-54478114 on Build GRCh38
rs753326203 Chr.14: 54478077-54478077 on Build GRCh38
rs759856873 Chr.14: 54478060-54478060 on Build GRCh38
rs773225455 Chr.14: 54478046-54478046 on Build GRCh38
rs376566387 Chr.14: 54478068-54478068 on Build GRCh38
rs755823430 Chr.14: 54478163-54478164 on Build GRCh38
rs774212905 Chr.14: 54478039-54478039 on Build GRCh38
rs922226203 Chr.14: 54478043-54478043 on Build GRCh38
rs749085724 Chr.14: 54478164-54478165 on Build GRCh38
rs866066642 Chr.14: 54478160-54478160 on Build GRCh38
rs762279031 Chr.14: 54478020-54478020 on Build GRCh38
rs942440670 Chr.14: 54478008-54478008 on Build GRCh38
rs767413305 Chr.14: 54478061-54478061 on Build GRCh38
rs777047523 Chr.14: 54478014-54478014 on Build GRCh38
rs369664579 Chr.14: 54478139-54478139 on Build GRCh38
rs76915130 Chr.14: 54478176-54478177 on Build GRCh38
rs976446573 Chr.14: 54478013-54478016 on Build GRCh38
rs377630752 Chr.14: 54478133-54478133 on Build GRCh38
rs752576949 Chr.14: 54478063-54478063 on Build GRCh38
rs545298017 Chr.14: 54478164-54478164 on Build GRCh38
rs906504732 Chr.14: 54478164-54478164 on Build GRCh38
rs373386829 Chr.14: 54478050-54478050 on Build GRCh38
rs200777099 Chr.14: 54478113-54478113 on Build GRCh38
rs756069684 Chr.14: 54478064-54478064 on Build GRCh38
rs911007831 Chr.14: 54478007-54478007 on Build GRCh38

View Full Product Details