Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93470956-93471225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120A
Entrez Gene ID
23196
Gene Name
family with sequence similarity 120A
Gene Aliases
C9orf10, OSSA
UniGene
Hs.372003
Gene Chromosome Location
Chr.9: 93451696-93566115 on Build GRCh38
Transcripts
NM_001286722.1 NM_001286723.1 NM_001286724.1 NM_014612.4 XM_005251842.4 XM_011518412.2 XM_011518413.2 XM_011518414.2 XM_011518416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909041 c.512T>C p.I171T Chr.9: 93471178-93471178 on Build GRCh38
3909039 c.512T>C p.I171T Chr.9: 93471178-93471178 on Build GRCh38
1463714 c.552G>A p.A184A Chr.9: 93471218-93471218 on Build GRCh38
5101497 c.552G>A p.A184A Chr.9: 93471218-93471218 on Build GRCh38
3909040 c.512T>C p.I171T Chr.9: 93471178-93471178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936783603 Chr.9: 93471145-93471145 on Build GRCh38
rs925743648 Chr.9: 93471066-93471066 on Build GRCh38
rs553257804 Chr.9: 93471132-93471132 on Build GRCh38
rs772023913 Chr.9: 93471155-93471155 on Build GRCh38
rs955854396 Chr.9: 93471167-93471167 on Build GRCh38
rs765334119 Chr.9: 93471040-93471040 on Build GRCh38
rs761650083 Chr.9: 93471203-93471203 on Build GRCh38
rs776687072 Chr.9: 93471199-93471199 on Build GRCh38
rs976896527 Chr.9: 93471196-93471196 on Build GRCh38
rs985293919 Chr.9: 93471031-93471031 on Build GRCh38
rs953901304 Chr.9: 93471029-93471029 on Build GRCh38
rs575302951 Chr.9: 93471107-93471107 on Build GRCh38
rs755467517 Chr.9: 93471093-93471093 on Build GRCh38
rs745985423 Chr.9: 93471154-93471154 on Build GRCh38
rs771230098 Chr.9: 93471133-93471133 on Build GRCh38
rs748593975 Chr.9: 93471105-93471105 on Build GRCh38
rs373865754 Chr.9: 93471131-93471131 on Build GRCh38
rs1029579546 Chr.9: 93471016-93471016 on Build GRCh38
rs976695851 Chr.9: 93471015-93471018 on Build GRCh38
rs781740999 Chr.9: 93471098-93471098 on Build GRCh38
rs765083612 Chr.9: 93471204-93471204 on Build GRCh38
rs139309200 Chr.9: 93471139-93471139 on Build GRCh38
rs778930147 Chr.9: 93471028-93471028 on Build GRCh38
rs773488933 Chr.9: 93471111-93471111 on Build GRCh38
rs1031531809 Chr.9: 93471157-93471157 on Build GRCh38

View Full Product Details