Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180614742-180614989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
UniGene
Hs.712820
Gene Chromosome Location
Chr.3: 180614008-180679495 on Build GRCh38
Transcripts
NM_181426.1 NM_001288582.1

Gene Symbol
TTC14
Entrez Gene ID
151613
Gene Name
tetratricopeptide repeat domain 14
Gene Aliases
DRDL5813, PRO19630
UniGene
Hs.43213
Gene Chromosome Location
Chr.3: 180602130-180618347 on Build GRCh38
Transcripts
NM_181426.1 NM_001288582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4935926 c.2767A>G p.R923G Chr.3: 180614980-180614980 on Build GRCh38
6230537 c.2759G>A p.S920N Chr.3: 180614988-180614988 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370902814 Chr.3: 180614900-180614900 on Build GRCh38
rs556950924 Chr.3: 180614944-180614945 on Build GRCh38
rs113696248 Chr.3: 180614788-180614788 on Build GRCh38
rs750798797 Chr.3: 180614885-180614885 on Build GRCh38
rs550082047 Chr.3: 180614820-180614820 on Build GRCh38
rs190913291 Chr.3: 180614787-180614787 on Build GRCh38
rs771010943 Chr.3: 180614963-180614963 on Build GRCh38
rs971432073 Chr.3: 180614869-180614869 on Build GRCh38
rs886058197 Chr.3: 180614947-180614947 on Build GRCh38
rs199649763 Chr.3: 180614952-180614952 on Build GRCh38
rs201549088 Chr.3: 180614788-180614789 on Build GRCh38
rs780168478 Chr.3: 180614914-180614914 on Build GRCh38
rs1007008869 Chr.3: 180614806-180614806 on Build GRCh38
rs957105046 Chr.3: 180614823-180614823 on Build GRCh38
rs1014569413 Chr.3: 180614822-180614822 on Build GRCh38
rs887714654 Chr.3: 180614789-180614789 on Build GRCh38
rs959085488 Chr.3: 180614887-180614887 on Build GRCh38
rs773632317 Chr.3: 180614774-180614774 on Build GRCh38
rs529451919 Chr.3: 180614856-180614856 on Build GRCh38
rs147012409 Chr.3: 180614846-180614846 on Build GRCh38
rs971110254 Chr.3: 180614827-180614827 on Build GRCh38
rs781387509 Chr.3: 180614936-180614936 on Build GRCh38
rs758739534 Chr.3: 180614892-180614892 on Build GRCh38
rs759348071 Chr.3: 180614950-180614952 on Build GRCh38
rs990339739 Chr.3: 180614927-180614927 on Build GRCh38
rs200947486 Chr.3: 180614930-180614930 on Build GRCh38
rs751844801 Chr.3: 180614924-180614924 on Build GRCh38

View Full Product Details