Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16890371-16890644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL3
Entrez Gene ID
9002
Gene Name
F2R like thrombin/trypsin receptor 3
Gene Aliases
PAR4
UniGene
Hs.137574
Gene Chromosome Location
Chr.19: 16889015-16892019 on Build GRCh38
Transcripts
NM_003950.2 XM_005260139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6228378 c.1079C>T p.P360L Chr.19: 16890542-16890542 on Build GRCh38
3530196 c.1044G>A p.R348R Chr.19: 16890507-16890507 on Build GRCh38
3530195 c.1036G>A p.E346K Chr.19: 16890499-16890499 on Build GRCh38
6302268 c.975C>T p.S325S Chr.19: 16890438-16890438 on Build GRCh38
6370018 c.1123C>T p.R375W Chr.19: 16890586-16890586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758078409 Chr.19: 16890443-16890443 on Build GRCh38
rs553086888 Chr.19: 16890465-16890465 on Build GRCh38
rs781055286 Chr.19: 16890393-16890393 on Build GRCh38
rs201593664 Chr.19: 16890403-16890403 on Build GRCh38
rs774604184 Chr.19: 16890409-16890409 on Build GRCh38
rs139232968 Chr.19: 16890466-16890466 on Build GRCh38
rs1019666833 Chr.19: 16890600-16890600 on Build GRCh38
rs750931051 Chr.19: 16890427-16890427 on Build GRCh38
rs369129094 Chr.19: 16890542-16890542 on Build GRCh38
rs145145785 Chr.19: 16890573-16890573 on Build GRCh38
rs773433039 Chr.19: 16890545-16890545 on Build GRCh38
rs775640062 Chr.19: 16890402-16890402 on Build GRCh38
rs745708136 Chr.19: 16890397-16890397 on Build GRCh38
rs776978440 Chr.19: 16890553-16890553 on Build GRCh38
rs184776893 Chr.19: 16890526-16890526 on Build GRCh38
rs11667441 Chr.19: 16890445-16890445 on Build GRCh38
rs768945179 Chr.19: 16890477-16890477 on Build GRCh38
rs550201673 Chr.19: 16890556-16890556 on Build GRCh38
rs767382226 Chr.19: 16890498-16890498 on Build GRCh38
rs772276253 Chr.19: 16890490-16890490 on Build GRCh38
rs771486970 Chr.19: 16890550-16890550 on Build GRCh38
rs768188622 Chr.19: 16890419-16890419 on Build GRCh38
rs778096365 Chr.19: 16890606-16890606 on Build GRCh38
rs371852294 Chr.19: 16890524-16890524 on Build GRCh38
rs758728803 Chr.19: 16890603-16890603 on Build GRCh38
rs747158588 Chr.19: 16890613-16890613 on Build GRCh38
rs761261251 Chr.19: 16890494-16890494 on Build GRCh38
rs777246611 Chr.19: 16890444-16890444 on Build GRCh38
rs530112267 Chr.19: 16890552-16890552 on Build GRCh38
rs111890288 Chr.19: 16890448-16890448 on Build GRCh38
rs747024311 Chr.19: 16890549-16890549 on Build GRCh38
rs142442138 Chr.19: 16890514-16890514 on Build GRCh38
rs1056228495 Chr.19: 16890513-16890513 on Build GRCh38
rs762024920 Chr.19: 16890489-16890489 on Build GRCh38
rs751089116 Chr.19: 16890447-16890447 on Build GRCh38
rs533771844 Chr.19: 16890622-16890622 on Build GRCh38
rs2227347 Chr.19: 16890429-16890429 on Build GRCh38
rs571332331 Chr.19: 16890589-16890589 on Build GRCh38
rs747888739 Chr.19: 16890601-16890601 on Build GRCh38
rs201379450 Chr.19: 16890509-16890509 on Build GRCh38
rs2227348 Chr.19: 16890543-16890543 on Build GRCh38
rs754699995 Chr.19: 16890540-16890540 on Build GRCh38
rs749158087 Chr.19: 16890536-16890536 on Build GRCh38
rs754862551 Chr.19: 16890591-16890591 on Build GRCh38
rs375412312 Chr.19: 16890411-16890411 on Build GRCh38
rs899275165 Chr.19: 16890391-16890391 on Build GRCh38
rs149415074 Chr.19: 16890439-16890439 on Build GRCh38
rs141455038 Chr.19: 16890572-16890572 on Build GRCh38
rs766755673 Chr.19: 16890430-16890430 on Build GRCh38
rs759981822 Chr.19: 16890555-16890555 on Build GRCh38
rs376378687 Chr.19: 16890597-16890597 on Build GRCh38
rs2227376 Chr.19: 16890392-16890392 on Build GRCh38
rs779807451 Chr.19: 16890535-16890535 on Build GRCh38
rs781145358 Chr.19: 16890461-16890461 on Build GRCh38
rs777089838 Chr.19: 16890495-16890495 on Build GRCh38
rs769597368 Chr.19: 16890400-16890400 on Build GRCh38
rs369983396 Chr.19: 16890522-16890522 on Build GRCh38
rs753717142 Chr.19: 16890587-16890587 on Build GRCh38
rs376656372 Chr.19: 16890518-16890518 on Build GRCh38
rs773916969 Chr.19: 16890492-16890492 on Build GRCh38
rs551525601 Chr.19: 16890578-16890578 on Build GRCh38
rs1049189337 Chr.19: 16890497-16890497 on Build GRCh38
rs774693812 Chr.19: 16890486-16890486 on Build GRCh38

View Full Product Details