Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27081367-27081598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf71
Entrez Gene ID
728858
Gene Name
chromosome 12 open reading frame 71
Gene Aliases
-
UniGene
Hs.334003
Gene Chromosome Location
Chr.12: 27081057-27084548 on Build GRCh38
Transcripts
NM_001080406.1 XM_011520847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666808 c.553G>A p.A185T Chr.12: 27081431-27081431 on Build GRCh38
348641 c.534C>A p.T178T Chr.12: 27081450-27081450 on Build GRCh38
6136440 c.598G>A p.D200N Chr.12: 27081386-27081386 on Build GRCh38
2203576 c.535G>A p.A179T Chr.12: 27081449-27081449 on Build GRCh38
5743546 c.617_618insCA p.Q206fs*7 Chr.12: 27081366-27081367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545680328 Chr.12: 27081526-27081526 on Build GRCh38
rs764897632 Chr.12: 27081451-27081451 on Build GRCh38
rs531548082 Chr.12: 27081450-27081450 on Build GRCh38
rs990576277 Chr.12: 27081535-27081535 on Build GRCh38
rs1023434579 Chr.12: 27081565-27081565 on Build GRCh38
rs772039371 Chr.12: 27081402-27081402 on Build GRCh38
rs751947864 Chr.12: 27081501-27081501 on Build GRCh38
rs34457591 Chr.12: 27081572-27081572 on Build GRCh38
rs952948850 Chr.12: 27081517-27081517 on Build GRCh38
rs768320743 Chr.12: 27081492-27081495 on Build GRCh38
rs556133776 Chr.12: 27081456-27081456 on Build GRCh38
rs757800284 Chr.12: 27081507-27081507 on Build GRCh38
rs189463694 Chr.12: 27081511-27081511 on Build GRCh38
rs202038202 Chr.12: 27081429-27081429 on Build GRCh38
rs764589837 Chr.12: 27081486-27081486 on Build GRCh38
rs1019602182 Chr.12: 27081447-27081447 on Build GRCh38
rs773007866 Chr.12: 27081406-27081406 on Build GRCh38
rs760038057 Chr.12: 27081483-27081483 on Build GRCh38
rs77584722 Chr.12: 27081431-27081431 on Build GRCh38
rs1049923351 Chr.12: 27081395-27081395 on Build GRCh38
rs377749308 Chr.12: 27081391-27081391 on Build GRCh38
rs747923429 Chr.12: 27081389-27081389 on Build GRCh38
rs539887044 Chr.12: 27081432-27081432 on Build GRCh38
rs759136904 Chr.12: 27081445-27081445 on Build GRCh38

View Full Product Details