Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179678877-179679132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANX
Entrez Gene ID
821
Gene Name
calnexin
Gene Aliases
CNX, IP90, P90
UniGene
Hs.567968
Gene Chromosome Location
Chr.5: 179678631-179731641 on Build GRCh38
Transcripts
XM_011534665.2 NM_001164444.1 XM_017009726.1

Gene Symbol
CBY3
Entrez Gene ID
646019
Gene Name
chibby family member 3
Gene Aliases
-
UniGene
Hs.535767
Gene Chromosome Location
Chr.5: 179677969-179681830 on Build GRCh38
Transcripts
XM_011534665.2 NM_001164444.1 XM_017009726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415455 c.193G>T p.A65S Chr.5: 179679119-179679119 on Build GRCh38
4155788 c.345G>A p.E115E Chr.5: 179678967-179678967 on Build GRCh38
6225234 c.364G>A p.A122T Chr.5: 179678948-179678948 on Build GRCh38
6286070 c.387C>A p.N129K Chr.5: 179678925-179678925 on Build GRCh38
7255016 c.427A>C p.S143R Chr.5: 179678885-179678885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911964920 Chr.5: 179678985-179678985 on Build GRCh38
rs34490996 Chr.5: 179678918-179678919 on Build GRCh38
rs755854814 Chr.5: 179679077-179679077 on Build GRCh38
rs748712355 Chr.5: 179678964-179678964 on Build GRCh38
rs750260749 Chr.5: 179678904-179678904 on Build GRCh38
rs937622709 Chr.5: 179679080-179679080 on Build GRCh38
rs954548673 Chr.5: 179678940-179678940 on Build GRCh38
rs751513335 Chr.5: 179678961-179678961 on Build GRCh38
rs1046150925 Chr.5: 179679110-179679110 on Build GRCh38
rs187808595 Chr.5: 179679101-179679101 on Build GRCh38
rs530037246 Chr.5: 179679068-179679068 on Build GRCh38
rs943403138 Chr.5: 179679030-179679030 on Build GRCh38
rs979192693 Chr.5: 179679005-179679005 on Build GRCh38
rs35217587 Chr.5: 179679073-179679074 on Build GRCh38
rs1056064556 Chr.5: 179679083-179679083 on Build GRCh38
rs1028750251 Chr.5: 179678939-179678939 on Build GRCh38
rs866548819 Chr.5: 179679114-179679114 on Build GRCh38
rs866896002 Chr.5: 179679092-179679092 on Build GRCh38
rs777528778 Chr.5: 179679078-179679078 on Build GRCh38
rs116305151 Chr.5: 179679095-179679095 on Build GRCh38
rs1047966904 Chr.5: 179679103-179679103 on Build GRCh38
rs913333521 Chr.5: 179678967-179678967 on Build GRCh38
rs531243590 Chr.5: 179678973-179678973 on Build GRCh38
rs762583894 Chr.5: 179678906-179678906 on Build GRCh38
rs750711526 Chr.5: 179678995-179679001 on Build GRCh38
rs563732089 Chr.5: 179678965-179678965 on Build GRCh38
rs767178290 Chr.5: 179679003-179679003 on Build GRCh38
rs751468879 Chr.5: 179678998-179678999 on Build GRCh38
rs760010939 Chr.5: 179679102-179679102 on Build GRCh38
rs1039234880 Chr.5: 179679033-179679033 on Build GRCh38
rs766357097 Chr.5: 179678924-179678924 on Build GRCh38
rs967394223 Chr.5: 179678983-179678983 on Build GRCh38
rs752235289 Chr.5: 179679025-179679025 on Build GRCh38
rs915051017 Chr.5: 179678895-179678895 on Build GRCh38
rs769343609 Chr.5: 179678968-179678968 on Build GRCh38

View Full Product Details