Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95177600-95177843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESN3
Entrez Gene ID
143686
Gene Name
sestrin 3
Gene Aliases
SEST3
UniGene
Hs.120633
Gene Chromosome Location
Chr.11: 95165513-95232541 on Build GRCh38
Transcripts
NM_001271594.1 NM_144665.3 XM_011542614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4037742 c.1229A>G p.H410R Chr.11: 95177737-95177737 on Build GRCh38
1659948 c.1160A>G p.Y387C Chr.11: 95177806-95177806 on Build GRCh38
3670925 c.1151A>G p.N384S Chr.11: 95177815-95177815 on Build GRCh38
3792060 c.1180G>A p.E394K Chr.11: 95177786-95177786 on Build GRCh38
4804531 c.1245C>T p.I415I Chr.11: 95177721-95177721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003818156 Chr.11: 95177662-95177662 on Build GRCh38
rs142438341 Chr.11: 95177805-95177805 on Build GRCh38
rs368390546 Chr.11: 95177710-95177710 on Build GRCh38
rs775518599 Chr.11: 95177784-95177784 on Build GRCh38
rs202245724 Chr.11: 95177763-95177763 on Build GRCh38
rs148408576 Chr.11: 95177769-95177769 on Build GRCh38
rs777309618 Chr.11: 95177756-95177756 on Build GRCh38
rs544818925 Chr.11: 95177637-95177637 on Build GRCh38
rs777032943 Chr.11: 95177700-95177700 on Build GRCh38
rs768817894 Chr.11: 95177789-95177789 on Build GRCh38
rs755803450 Chr.11: 95177746-95177746 on Build GRCh38
rs971920149 Chr.11: 95177704-95177704 on Build GRCh38
rs765736032 Chr.11: 95177707-95177707 on Build GRCh38
rs766049473 Chr.11: 95177699-95177699 on Build GRCh38
rs752292702 Chr.11: 95177736-95177736 on Build GRCh38
rs747143626 Chr.11: 95177787-95177787 on Build GRCh38
rs112153772 Chr.11: 95177668-95177668 on Build GRCh38
rs145434950 Chr.11: 95177762-95177762 on Build GRCh38
rs146666544 Chr.11: 95177701-95177701 on Build GRCh38
rs542742332 Chr.11: 95177714-95177714 on Build GRCh38
rs749067368 Chr.11: 95177761-95177761 on Build GRCh38
rs764341735 Chr.11: 95177687-95177687 on Build GRCh38
rs866735127 Chr.11: 95177642-95177642 on Build GRCh38
rs760310951 Chr.11: 95177691-95177692 on Build GRCh38
rs988594861 Chr.11: 95177726-95177726 on Build GRCh38
rs958679635 Chr.11: 95177806-95177806 on Build GRCh38
rs767125203 Chr.11: 95177720-95177720 on Build GRCh38
rs772062473 Chr.11: 95177770-95177770 on Build GRCh38
rs760880661 Chr.11: 95177672-95177672 on Build GRCh38
rs776751070 Chr.11: 95177798-95177798 on Build GRCh38
rs563016595 Chr.11: 95177644-95177644 on Build GRCh38
rs910664812 Chr.11: 95177782-95177782 on Build GRCh38
rs200790229 Chr.11: 95177735-95177735 on Build GRCh38
rs757329819 Chr.11: 95177702-95177702 on Build GRCh38

View Full Product Details