Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37909538-37909797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFRP4
Entrez Gene ID
6424
Gene Name
secreted frizzled related protein 4
Gene Aliases
FRP-4, FRPHE, sFRP-4
UniGene
Hs.658169
Gene Chromosome Location
Chr.7: 37905932-37916923 on Build GRCh38
Transcripts
NM_003014.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1265391 c.792-4_792-3insT p.? Chr.7: 37909683-37909684 on Build GRCh38
4559471 c.805G>A p.E269K Chr.7: 37909667-37909667 on Build GRCh38
6776405 c.800T>G p.L267R Chr.7: 37909672-37909672 on Build GRCh38
485301 c.798G>T p.M266I Chr.7: 37909674-37909674 on Build GRCh38
6204510 c.792-3C>T p.? Chr.7: 37909683-37909683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753245114 Chr.7: 37909651-37909651 on Build GRCh38
rs778931657 Chr.7: 37909709-37909709 on Build GRCh38
rs750708442 Chr.7: 37909690-37909690 on Build GRCh38
rs186831726 Chr.7: 37909716-37909716 on Build GRCh38
rs771759619 Chr.7: 37909614-37909614 on Build GRCh38
rs905630779 Chr.7: 37909756-37909756 on Build GRCh38
rs749370781 Chr.7: 37909705-37909705 on Build GRCh38
rs868460292 Chr.7: 37909625-37909625 on Build GRCh38
rs756170366 Chr.7: 37909695-37909695 on Build GRCh38
rs752782152 Chr.7: 37909588-37909588 on Build GRCh38
rs766055041 Chr.7: 37909633-37909633 on Build GRCh38
rs773046051 Chr.7: 37909623-37909623 on Build GRCh38
rs1001676289 Chr.7: 37909758-37909758 on Build GRCh38
rs760229761 Chr.7: 37909624-37909624 on Build GRCh38
rs771940642 Chr.7: 37909719-37909719 on Build GRCh38
rs764670979 Chr.7: 37909670-37909670 on Build GRCh38
rs767461457 Chr.7: 37909572-37909572 on Build GRCh38
rs764061976 Chr.7: 37909698-37909698 on Build GRCh38
rs553024916 Chr.7: 37909720-37909720 on Build GRCh38
rs1013928157 Chr.7: 37909747-37909747 on Build GRCh38
rs147388554 Chr.7: 37909680-37909680 on Build GRCh38
rs749279302 Chr.7: 37909608-37909608 on Build GRCh38
rs763497705 Chr.7: 37909668-37909668 on Build GRCh38
rs768648647 Chr.7: 37909611-37909611 on Build GRCh38
rs941263462 Chr.7: 37909615-37909615 on Build GRCh38
rs903450219 Chr.7: 37909757-37909757 on Build GRCh38
rs751834775 Chr.7: 37909673-37909673 on Build GRCh38
rs773948148 Chr.7: 37909612-37909612 on Build GRCh38
rs754944705 Chr.7: 37909590-37909590 on Build GRCh38
rs999170498 Chr.7: 37909775-37909775 on Build GRCh38
rs745763643 Chr.7: 37909684-37909684 on Build GRCh38
rs144196534 Chr.7: 37909679-37909679 on Build GRCh38
rs947584055 Chr.7: 37909743-37909743 on Build GRCh38
rs1039803036 Chr.7: 37909708-37909708 on Build GRCh38
rs780669813 Chr.7: 37909621-37909621 on Build GRCh38
rs772996708 Chr.7: 37909726-37909726 on Build GRCh38
rs760464463 Chr.7: 37909729-37909729 on Build GRCh38
rs372476594 Chr.7: 37909569-37909569 on Build GRCh38
rs769395024 Chr.7: 37909683-37909684 on Build GRCh38
rs896803900 Chr.7: 37909692-37909692 on Build GRCh38
rs1045850803 Chr.7: 37909748-37909748 on Build GRCh38
rs761698667 Chr.7: 37909613-37909613 on Build GRCh38

View Full Product Details