Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44069027-44069298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARVB
Entrez Gene ID
29780
Gene Name
parvin beta
Gene Aliases
CGI-56
UniGene
Hs.475074
Gene Chromosome Location
Chr.22: 43999211-44169233 on Build GRCh38
Transcripts
NM_001003828.2 NM_001243385.1 NM_001243386.1 NM_013327.4 XM_005261596.1 XM_017028792.1

SNP Detail

SNP ID Chromosome Location
rs536646235 Chr.22: 44069150-44069150 on Build GRCh38
rs189322999 Chr.22: 44069058-44069058 on Build GRCh38
rs373273192 Chr.22: 44069170-44069170 on Build GRCh38
rs537272848 Chr.22: 44069105-44069105 on Build GRCh38
rs1021220210 Chr.22: 44069262-44069262 on Build GRCh38
rs114129729 Chr.22: 44069081-44069081 on Build GRCh38
rs768673285 Chr.22: 44069136-44069136 on Build GRCh38
rs761683371 Chr.22: 44069089-44069089 on Build GRCh38
rs557411336 Chr.22: 44069108-44069108 on Build GRCh38
rs1049432863 Chr.22: 44069183-44069183 on Build GRCh38
rs567693425 Chr.22: 44069113-44069113 on Build GRCh38
rs551568673 Chr.22: 44069082-44069082 on Build GRCh38
rs769567134 Chr.22: 44069145-44069145 on Build GRCh38
rs565922225 Chr.22: 44069068-44069068 on Build GRCh38
rs369853280 Chr.22: 44069140-44069140 on Build GRCh38
rs779626714 Chr.22: 44069120-44069120 on Build GRCh38
rs959466887 Chr.22: 44069118-44069118 on Build GRCh38
rs771779713 Chr.22: 44069116-44069116 on Build GRCh38
rs768117158 Chr.22: 44069164-44069164 on Build GRCh38
rs898212222 Chr.22: 44069060-44069060 on Build GRCh38
rs746452034 Chr.22: 44069130-44069130 on Build GRCh38
rs1004249498 Chr.22: 44069056-44069056 on Build GRCh38
rs1008440486 Chr.22: 44069086-44069086 on Build GRCh38
rs761649214 Chr.22: 44069141-44069141 on Build GRCh38
rs778282382 Chr.22: 44069181-44069181 on Build GRCh38
rs756610147 Chr.22: 44069180-44069180 on Build GRCh38
rs137958680 Chr.22: 44069158-44069158 on Build GRCh38
rs763166394 Chr.22: 44069154-44069154 on Build GRCh38
rs756524443 Chr.22: 44069112-44069112 on Build GRCh38
rs558697154 Chr.22: 44069221-44069221 on Build GRCh38
rs758088597 Chr.22: 44069206-44069206 on Build GRCh38
rs968219356 Chr.22: 44069272-44069272 on Build GRCh38
rs895208732 Chr.22: 44069186-44069186 on Build GRCh38
rs73888833 Chr.22: 44069156-44069156 on Build GRCh38
rs754195544 Chr.22: 44069198-44069198 on Build GRCh38
rs766008907 Chr.22: 44069166-44069169 on Build GRCh38
rs745507196 Chr.22: 44069115-44069115 on Build GRCh38
rs1022405782 Chr.22: 44069266-44069266 on Build GRCh38
rs370885326 Chr.22: 44069159-44069159 on Build GRCh38
rs955530189 Chr.22: 44069084-44069084 on Build GRCh38
rs571489747 Chr.22: 44069094-44069094 on Build GRCh38
rs891294755 Chr.22: 44069219-44069219 on Build GRCh38

View Full Product Details