Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99550772-99551031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT10A
Entrez Gene ID
93587
Gene Name
tRNA methyltransferase 10A
Gene Aliases
HEL-S-88, MSSGM, MSSGM1, RG9MTD2, TRM10
UniGene
Hs.380412
Gene Chromosome Location
Chr.4: 99546707-99564057 on Build GRCh38
Transcripts
NM_001134665.2 NM_001134666.2 NM_152292.4 XM_005263352.3 XM_006714417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844472 c.668C>T p.S223L Chr.4: 99550968-99550968 on Build GRCh38
6808955 c.665C>T p.A222V Chr.4: 99550971-99550971 on Build GRCh38
4945185 c.693G>T p.Q231H Chr.4: 99550943-99550943 on Build GRCh38
3824827 c.717G>C p.K239N Chr.4: 99550919-99550919 on Build GRCh38
1049429 c.728G>A p.R243Q Chr.4: 99550908-99550908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767830309 Chr.4: 99550886-99550886 on Build GRCh38
rs748618515 Chr.4: 99550989-99550989 on Build GRCh38
rs765351342 Chr.4: 99550916-99550916 on Build GRCh38
rs752632280 Chr.4: 99550918-99550918 on Build GRCh38
rs898944165 Chr.4: 99550803-99550803 on Build GRCh38
rs758295087 Chr.4: 99550947-99550947 on Build GRCh38
rs746764425 Chr.4: 99550959-99550959 on Build GRCh38
rs368697138 Chr.4: 99550912-99550912 on Build GRCh38
rs371843840 Chr.4: 99550985-99550985 on Build GRCh38
rs576484676 Chr.4: 99550851-99550851 on Build GRCh38
rs137989992 Chr.4: 99550953-99550953 on Build GRCh38
rs868525420 Chr.4: 99550927-99550927 on Build GRCh38
rs770502540 Chr.4: 99550854-99550854 on Build GRCh38
rs776150087 Chr.4: 99550855-99550855 on Build GRCh38
rs145480253 Chr.4: 99550908-99550908 on Build GRCh38
rs755001904 Chr.4: 99550844-99550844 on Build GRCh38
rs375250044 Chr.4: 99550842-99550842 on Build GRCh38
rs753997546 Chr.4: 99550911-99550911 on Build GRCh38
rs375746140 Chr.4: 99550849-99550849 on Build GRCh38
rs755903549 Chr.4: 99550943-99550943 on Build GRCh38
rs994542092 Chr.4: 99550813-99550813 on Build GRCh38
rs371814008 Chr.4: 99550848-99550848 on Build GRCh38
rs745332389 Chr.4: 99550867-99550867 on Build GRCh38
rs774760866 Chr.4: 99550877-99550877 on Build GRCh38
rs142490083 Chr.4: 99550966-99550966 on Build GRCh38
rs756924083 Chr.4: 99550962-99550962 on Build GRCh38
rs13129571 Chr.4: 99550847-99550847 on Build GRCh38
rs771825867 Chr.4: 99550850-99550850 on Build GRCh38
rs140905333 Chr.4: 99550971-99550971 on Build GRCh38
rs1043513557 Chr.4: 99550879-99550879 on Build GRCh38
rs762378878 Chr.4: 99550878-99550878 on Build GRCh38
rs745489199 Chr.4: 99550970-99550970 on Build GRCh38
rs755048644 Chr.4: 99550914-99550914 on Build GRCh38
rs769325542 Chr.4: 99550868-99550868 on Build GRCh38
rs773655792 Chr.4: 99550892-99550892 on Build GRCh38

View Full Product Details