Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62960563-62960814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
UniGene
Hs.512686
Gene Chromosome Location
Chr.20: 62952647-62968597 on Build GRCh38
Transcripts
NM_001302643.1 NM_022082.3 XM_011528978.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5577050 c.459C>A p.A153A Chr.20: 62960565-62960565 on Build GRCh38
399429 c.489C>T p.S163S Chr.20: 62960595-62960595 on Build GRCh38
4618061 c.466T>C p.Y156H Chr.20: 62960572-62960572 on Build GRCh38
6572893 c.472A>G p.I158V Chr.20: 62960578-62960578 on Build GRCh38
1681839 c.484G>A p.G162S Chr.20: 62960590-62960590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748882560 Chr.20: 62960599-62960599 on Build GRCh38
rs745835085 Chr.20: 62960625-62960625 on Build GRCh38
rs1028225295 Chr.20: 62960623-62960623 on Build GRCh38
rs530769465 Chr.20: 62960742-62960743 on Build GRCh38
rs755635858 Chr.20: 62960596-62960596 on Build GRCh38
rs967923285 Chr.20: 62960610-62960610 on Build GRCh38
rs112060558 Chr.20: 62960735-62960735 on Build GRCh38
rs1009957927 Chr.20: 62960786-62960786 on Build GRCh38
rs777580284 Chr.20: 62960598-62960598 on Build GRCh38
rs928204519 Chr.20: 62960780-62960780 on Build GRCh38
rs770915034 Chr.20: 62960611-62960611 on Build GRCh38
rs919642102 Chr.20: 62960683-62960683 on Build GRCh38
rs913003536 Chr.20: 62960792-62960792 on Build GRCh38
rs996658440 Chr.20: 62960791-62960791 on Build GRCh38
rs951451079 Chr.20: 62960713-62960713 on Build GRCh38
rs147701965 Chr.20: 62960790-62960790 on Build GRCh38
rs112626678 Chr.20: 62960730-62960730 on Build GRCh38
rs772079523 Chr.20: 62960641-62960641 on Build GRCh38
rs752279300 Chr.20: 62960586-62960586 on Build GRCh38
rs970052844 Chr.20: 62960729-62960729 on Build GRCh38
rs1022389838 Chr.20: 62960789-62960789 on Build GRCh38
rs1017030508 Chr.20: 62960726-62960726 on Build GRCh38
rs375386260 Chr.20: 62960647-62960647 on Build GRCh38
rs1025711314 Chr.20: 62960676-62960676 on Build GRCh38
rs776078093 Chr.20: 62960639-62960640 on Build GRCh38
rs1045241954 Chr.20: 62960728-62960728 on Build GRCh38
rs955272981 Chr.20: 62960637-62960637 on Build GRCh38
rs938394946 Chr.20: 62960783-62960783 on Build GRCh38
rs1056694983 Chr.20: 62960760-62960760 on Build GRCh38
rs998583105 Chr.20: 62960627-62960627 on Build GRCh38
rs973868782 Chr.20: 62960669-62960669 on Build GRCh38
rs939644439 Chr.20: 62960665-62960665 on Build GRCh38
rs761898583 Chr.20: 62960648-62960648 on Build GRCh38
rs530633611 Chr.20: 62960698-62960698 on Build GRCh38
rs778749599 Chr.20: 62960616-62960616 on Build GRCh38
rs984273791 Chr.20: 62960720-62960720 on Build GRCh38

View Full Product Details