Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17259437-17259710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI3
Entrez Gene ID
51702
Gene Name
peptidyl arginine deiminase 3
Gene Aliases
PAD3, PDI3
UniGene
Hs.149195
Gene Chromosome Location
Chr.1: 17249079-17284233 on Build GRCh38
Transcripts
NM_016233.2 XM_011541571.2 XM_011541572.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736872 c.135G>A p.T45T Chr.1: 17259620-17259620 on Build GRCh38
3746668 c.154A>G p.I52V Chr.1: 17259639-17259639 on Build GRCh38
3966055 c.194C>A p.T65N Chr.1: 17259679-17259679 on Build GRCh38
899944 c.134C>T p.T45M Chr.1: 17259619-17259619 on Build GRCh38
6736870 c.160C>T p.P54S Chr.1: 17259645-17259645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375533169 Chr.1: 17259626-17259626 on Build GRCh38
rs765620074 Chr.1: 17259622-17259622 on Build GRCh38
rs775789894 Chr.1: 17259579-17259579 on Build GRCh38
rs1045777011 Chr.1: 17259509-17259509 on Build GRCh38
rs45438395 Chr.1: 17259515-17259515 on Build GRCh38
rs749231446 Chr.1: 17259552-17259552 on Build GRCh38
rs762348389 Chr.1: 17259620-17259620 on Build GRCh38
rs762034500 Chr.1: 17259678-17259678 on Build GRCh38
rs372937399 Chr.1: 17259548-17259548 on Build GRCh38
rs199924791 Chr.1: 17259619-17259619 on Build GRCh38
rs201025868 Chr.1: 17259560-17259560 on Build GRCh38
rs755819974 Chr.1: 17259651-17259651 on Build GRCh38
rs555384051 Chr.1: 17259547-17259547 on Build GRCh38
rs1001769596 Chr.1: 17259518-17259518 on Build GRCh38
rs376978164 Chr.1: 17259527-17259527 on Build GRCh38
rs764540424 Chr.1: 17259601-17259601 on Build GRCh38
rs3750300 Chr.1: 17259639-17259639 on Build GRCh38
rs201808059 Chr.1: 17259555-17259555 on Build GRCh38
rs113960275 Chr.1: 17259546-17259546 on Build GRCh38
rs148271347 Chr.1: 17259690-17259690 on Build GRCh38
rs553206163 Chr.1: 17259503-17259503 on Build GRCh38
rs772274412 Chr.1: 17259665-17259665 on Build GRCh38
rs780773147 Chr.1: 17259540-17259540 on Build GRCh38
rs772210276 Chr.1: 17259573-17259573 on Build GRCh38
rs761047531 Chr.1: 17259584-17259584 on Build GRCh38
rs745888593 Chr.1: 17259569-17259569 on Build GRCh38
rs201457466 Chr.1: 17259623-17259623 on Build GRCh38
rs150695491 Chr.1: 17259627-17259627 on Build GRCh38
rs199540947 Chr.1: 17259670-17259670 on Build GRCh38
rs891879574 Chr.1: 17259490-17259490 on Build GRCh38
rs111914086 Chr.1: 17259684-17259684 on Build GRCh38
rs539218252 Chr.1: 17259485-17259485 on Build GRCh38
rs200989099 Chr.1: 17259663-17259663 on Build GRCh38
rs770207385 Chr.1: 17259679-17259679 on Build GRCh38
rs115356766 Chr.1: 17259664-17259664 on Build GRCh38
rs146472416 Chr.1: 17259652-17259652 on Build GRCh38
rs747808353 Chr.1: 17259542-17259542 on Build GRCh38
rs142555354 Chr.1: 17259669-17259669 on Build GRCh38
rs974621772 Chr.1: 17259658-17259658 on Build GRCh38
rs1046441699 Chr.1: 17259491-17259491 on Build GRCh38
rs772200941 Chr.1: 17259502-17259502 on Build GRCh38
rs757100594 Chr.1: 17259659-17259659 on Build GRCh38
rs780338673 Chr.1: 17259666-17259666 on Build GRCh38
rs150292444 Chr.1: 17259691-17259691 on Build GRCh38
rs535785957 Chr.1: 17259531-17259531 on Build GRCh38
rs548507629 Chr.1: 17259672-17259672 on Build GRCh38
rs754545693 Chr.1: 17259537-17259537 on Build GRCh38
rs750962510 Chr.1: 17259625-17259625 on Build GRCh38

View Full Product Details