Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163505626-163505853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAT2B
Entrez Gene ID
27430
Gene Name
methionine adenosyltransferase 2B
Gene Aliases
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR
UniGene
Hs.54642
Gene Chromosome Location
Chr.5: 163503064-163519353 on Build GRCh38
Transcripts
NM_013283.4 NM_182796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286776 c.26C>G p.S9C Chr.5: 163505712-163505712 on Build GRCh38
6329971 c.7G>A p.G3R Chr.5: 163505693-163505693 on Build GRCh38
3410091 c.15G>A p.E5E Chr.5: 163505701-163505701 on Build GRCh38
3393402 c.9G>C p.G3G Chr.5: 163505695-163505695 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004688606 Chr.5: 163505824-163505824 on Build GRCh38
rs926988358 Chr.5: 163505646-163505646 on Build GRCh38
rs1034790776 Chr.5: 163505795-163505795 on Build GRCh38
rs746026958 Chr.5: 163505654-163505654 on Build GRCh38
rs764787880 Chr.5: 163505665-163505665 on Build GRCh38
rs764079729 Chr.5: 163505791-163505791 on Build GRCh38
rs944688744 Chr.5: 163505647-163505647 on Build GRCh38
rs759306922 Chr.5: 163505682-163505682 on Build GRCh38
rs762515711 Chr.5: 163505731-163505731 on Build GRCh38
rs901166851 Chr.5: 163505789-163505789 on Build GRCh38
rs752592788 Chr.5: 163505683-163505683 on Build GRCh38
rs760539375 Chr.5: 163505785-163505785 on Build GRCh38
rs762441416 Chr.5: 163505676-163505676 on Build GRCh38
rs998633634 Chr.5: 163505806-163505806 on Build GRCh38
rs543874089 Chr.5: 163505726-163505726 on Build GRCh38
rs299299 Chr.5: 163505816-163505816 on Build GRCh38
rs372087171 Chr.5: 163505658-163505658 on Build GRCh38
rs752645771 Chr.5: 163505784-163505784 on Build GRCh38
rs780300465 Chr.5: 163505710-163505710 on Build GRCh38
rs748604806 Chr.5: 163505723-163505723 on Build GRCh38
rs779225095 Chr.5: 163505653-163505653 on Build GRCh38
rs751219325 Chr.5: 163505651-163505651 on Build GRCh38
rs769336749 Chr.5: 163505659-163505659 on Build GRCh38
rs888045785 Chr.5: 163505697-163505697 on Build GRCh38
rs958588038 Chr.5: 163505751-163505751 on Build GRCh38
rs771798415 Chr.5: 163505719-163505719 on Build GRCh38
rs71599703 Chr.5: 163505747-163505747 on Build GRCh38
rs398088921 Chr.5: 163505800-163505800 on Build GRCh38
rs956618536 Chr.5: 163505809-163505809 on Build GRCh38
rs761436765 Chr.5: 163505662-163505662 on Build GRCh38
rs369711301 Chr.5: 163505681-163505681 on Build GRCh38
rs886384263 Chr.5: 163505823-163505823 on Build GRCh38
rs766055743 Chr.5: 163505678-163505678 on Build GRCh38
rs1040077237 Chr.5: 163505787-163505787 on Build GRCh38
rs555495270 Chr.5: 163505712-163505712 on Build GRCh38
rs758605300 Chr.5: 163505706-163505706 on Build GRCh38
rs79798488 Chr.5: 163505796-163505796 on Build GRCh38
rs938894796 Chr.5: 163505648-163505648 on Build GRCh38
rs774027377 Chr.5: 163505748-163505748 on Build GRCh38
rs759359935 Chr.5: 163505758-163505758 on Build GRCh38
rs904594076 Chr.5: 163505684-163505684 on Build GRCh38
rs199974025 Chr.5: 163505714-163505714 on Build GRCh38
rs750516631 Chr.5: 163505799-163505799 on Build GRCh38
rs760784149 Chr.5: 163505686-163505686 on Build GRCh38
rs767253204 Chr.5: 163505783-163505783 on Build GRCh38
rs777248803 Chr.5: 163505660-163505660 on Build GRCh38
rs750662270 Chr.5: 163505702-163505702 on Build GRCh38
rs559896861 Chr.5: 163505829-163505829 on Build GRCh38
rs757293921 Chr.5: 163505644-163505644 on Build GRCh38
rs577352717 Chr.5: 163505778-163505778 on Build GRCh38
rs1040463255 Chr.5: 163505650-163505650 on Build GRCh38
rs757351247 Chr.5: 163505688-163505688 on Build GRCh38
rs770542074 Chr.5: 163505745-163505745 on Build GRCh38
rs200516708 Chr.5: 163505657-163505657 on Build GRCh38
rs375373757 Chr.5: 163505668-163505668 on Build GRCh38
rs562221499 Chr.5: 163505728-163505728 on Build GRCh38
rs367719052 Chr.5: 163505643-163505643 on Build GRCh38
rs781769561 Chr.5: 163505717-163505717 on Build GRCh38
rs778802174 Chr.5: 163505689-163505689 on Build GRCh38

View Full Product Details