Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41421478-41421711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT37
Entrez Gene ID
8688
Gene Name
keratin 37
Gene Aliases
HA7, K37, KRTHA7
UniGene
Hs.673852
Gene Chromosome Location
Chr.17: 41420557-41424570 on Build GRCh38
Transcripts
NM_003770.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533539 c.1072G>A p.E358K Chr.17: 41421536-41421536 on Build GRCh38
6698017 c.1129C>T p.R377W Chr.17: 41421479-41421479 on Build GRCh38
6839390 c.1081C>T p.Q361* Chr.17: 41421527-41421527 on Build GRCh38
7198408 c.1024G>T p.D342Y Chr.17: 41421584-41421584 on Build GRCh38
5598438 c.1041C>T p.S347S Chr.17: 41421567-41421567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774073737 Chr.17: 41421541-41421541 on Build GRCh38
rs780920832 Chr.17: 41421510-41421510 on Build GRCh38
rs774198263 Chr.17: 41421624-41421624 on Build GRCh38
rs76042802 Chr.17: 41421543-41421543 on Build GRCh38
rs2857626 Chr.17: 41421632-41421632 on Build GRCh38
rs761563200 Chr.17: 41421633-41421633 on Build GRCh38
rs773166012 Chr.17: 41421638-41421638 on Build GRCh38
rs748902763 Chr.17: 41421620-41421620 on Build GRCh38
rs758590058 Chr.17: 41421556-41421556 on Build GRCh38
rs761796405 Chr.17: 41421542-41421542 on Build GRCh38
rs747531928 Chr.17: 41421503-41421503 on Build GRCh38
rs555741797 Chr.17: 41421669-41421669 on Build GRCh38
rs751971954 Chr.17: 41421557-41421557 on Build GRCh38
rs746325591 Chr.17: 41421582-41421582 on Build GRCh38
rs138622207 Chr.17: 41421555-41421555 on Build GRCh38
rs757881230 Chr.17: 41421564-41421564 on Build GRCh38
rs769215685 Chr.17: 41421594-41421594 on Build GRCh38
rs750578909 Chr.17: 41421501-41421501 on Build GRCh38
rs374599929 Chr.17: 41421636-41421636 on Build GRCh38
rs769531957 Chr.17: 41421515-41421515 on Build GRCh38
rs753958399 Chr.17: 41421545-41421545 on Build GRCh38
rs377623576 Chr.17: 41421623-41421623 on Build GRCh38
rs182995882 Chr.17: 41421534-41421534 on Build GRCh38
rs765322644 Chr.17: 41421548-41421548 on Build GRCh38
rs372151417 Chr.17: 41421528-41421528 on Build GRCh38
rs759557817 Chr.17: 41421547-41421547 on Build GRCh38
rs539555306 Chr.17: 41421504-41421504 on Build GRCh38
rs557914406 Chr.17: 41421512-41421512 on Build GRCh38
rs781759759 Chr.17: 41421574-41421574 on Build GRCh38
rs763768743 Chr.17: 41421585-41421587 on Build GRCh38
rs1016556000 Chr.17: 41421502-41421502 on Build GRCh38
rs77106064 Chr.17: 41421685-41421685 on Build GRCh38
rs373672102 Chr.17: 41421616-41421616 on Build GRCh38
rs775024542 Chr.17: 41421517-41421517 on Build GRCh38
rs916385074 Chr.17: 41421650-41421650 on Build GRCh38

View Full Product Details