Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104504671-104504940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR13F1
Entrez Gene ID
138805
Gene Name
olfactory receptor family 13 subfamily F member 1
Gene Aliases
OR9-6
UniGene
Hs.553602
Gene Chromosome Location
Chr.9: 104504263-104505222 on Build GRCh38
Transcripts
NM_001004485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5667795 c.629C>A p.P210Q Chr.9: 104504891-104504891 on Build GRCh38
1732792 c.558A>T p.K186N Chr.9: 104504820-104504820 on Build GRCh38
3652475 c.447G>A p.W149* Chr.9: 104504709-104504709 on Build GRCh38
5721368 c.675C>T p.I225I Chr.9: 104504937-104504937 on Build GRCh38
4709662 c.441C>A p.G147G Chr.9: 104504703-104504703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918511121 Chr.9: 104504699-104504699 on Build GRCh38
rs868203542 Chr.9: 104504800-104504800 on Build GRCh38
rs763390575 Chr.9: 104504727-104504727 on Build GRCh38
rs200785384 Chr.9: 104504864-104504864 on Build GRCh38
rs773728032 Chr.9: 104504721-104504721 on Build GRCh38
rs753296860 Chr.9: 104504880-104504880 on Build GRCh38
rs906247614 Chr.9: 104504906-104504906 on Build GRCh38
rs757160293 Chr.9: 104504787-104504787 on Build GRCh38
rs140294841 Chr.9: 104504837-104504837 on Build GRCh38
rs752398433 Chr.9: 104504768-104504768 on Build GRCh38
rs773177450 Chr.9: 104504832-104504832 on Build GRCh38
rs531765643 Chr.9: 104504805-104504805 on Build GRCh38
rs755620448 Chr.9: 104504770-104504770 on Build GRCh38
rs1012065671 Chr.9: 104504801-104504801 on Build GRCh38
rs781530157 Chr.9: 104504822-104504822 on Build GRCh38
rs780015091 Chr.9: 104504910-104504910 on Build GRCh38
rs759873030 Chr.9: 104504863-104504863 on Build GRCh38
rs760270925 Chr.9: 104504871-104504871 on Build GRCh38
rs79435118 Chr.9: 104504827-104504827 on Build GRCh38
rs372293721 Chr.9: 104504882-104504882 on Build GRCh38
rs267602065 Chr.9: 104504817-104504817 on Build GRCh38
rs778787214 Chr.9: 104504796-104504796 on Build GRCh38
rs761699519 Chr.9: 104504875-104504877 on Build GRCh38
rs201893993 Chr.9: 104504883-104504883 on Build GRCh38
rs766708470 Chr.9: 104504731-104504731 on Build GRCh38
rs767144728 Chr.9: 104504762-104504762 on Build GRCh38
rs551863082 Chr.9: 104504808-104504808 on Build GRCh38
rs542846013 Chr.9: 104504717-104504717 on Build GRCh38
rs796248802 Chr.9: 104504744-104504744 on Build GRCh38
rs752357086 Chr.9: 104504866-104504866 on Build GRCh38
rs145392760 Chr.9: 104504893-104504893 on Build GRCh38
rs748348638 Chr.9: 104504823-104504823 on Build GRCh38
rs1049057291 Chr.9: 104504873-104504873 on Build GRCh38
rs769861931 Chr.9: 104504825-104504825 on Build GRCh38
rs200699312 Chr.9: 104504777-104504777 on Build GRCh38
rs748343456 Chr.9: 104504915-104504915 on Build GRCh38
rs888264295 Chr.9: 104504772-104504772 on Build GRCh38
rs769911581 Chr.9: 104504916-104504916 on Build GRCh38
rs199737258 Chr.9: 104504858-104504858 on Build GRCh38
rs771424446 Chr.9: 104504850-104504850 on Build GRCh38
rs189551257 Chr.9: 104504902-104504902 on Build GRCh38
rs758435659 Chr.9: 104504807-104504807 on Build GRCh38
rs910445191 Chr.9: 104504734-104504734 on Build GRCh38
rs200767689 Chr.9: 104504847-104504847 on Build GRCh38
rs763604865 Chr.9: 104504879-104504879 on Build GRCh38
rs375969548 Chr.9: 104504786-104504786 on Build GRCh38
rs1048288174 Chr.9: 104504736-104504736 on Build GRCh38
rs774438612 Chr.9: 104504810-104504810 on Build GRCh38
rs761927781 Chr.9: 104504704-104504704 on Build GRCh38
rs750422191 Chr.9: 104504898-104504898 on Build GRCh38
rs368331596 Chr.9: 104504809-104504809 on Build GRCh38
rs759960983 Chr.9: 104504733-104504733 on Build GRCh38

View Full Product Details