Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20568428-20568701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED15
Entrez Gene ID
51586
Gene Name
mediator complex subunit 15
Gene Aliases
ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
UniGene
Hs.517421
Gene Chromosome Location
Chr.22: 20507542-20587632 on Build GRCh38
Transcripts
NM_001003891.2 NM_001293234.1 NM_001293235.1 NM_001293236.1 NM_001293237.1 NM_015889.4 XM_006724263.3 XM_006724264.3 XM_011530214.2 XM_011530215.2 XM_011530218.2 XM_011530219.1 XM_011530220.2 XM_017028817.1 XM_017028818.1 XM_017028819.1 XM_017028820.1 XM_017028821.1 XM_017028822.1 XM_017028823.1 XM_017028824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471404 c.1113G>C p.Q371H Chr.22: 20568592-20568592 on Build GRCh38
5565710 c.1046G>A p.R349Q Chr.22: 20568525-20568525 on Build GRCh38
5281618 c.1089_1091delGCA p.Q367delQ Chr.22: 20568568-20568570 on Build GRCh38
6506208 c.1087G>A p.V363M Chr.22: 20568566-20568566 on Build GRCh38
5565709 c.1046G>A p.R349Q Chr.22: 20568525-20568525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752939925 Chr.22: 20568637-20568637 on Build GRCh38
rs765298988 Chr.22: 20568517-20568517 on Build GRCh38
rs377387924 Chr.22: 20568675-20568675 on Build GRCh38
rs202216917 Chr.22: 20568623-20568623 on Build GRCh38
rs771734946 Chr.22: 20568592-20568592 on Build GRCh38
rs373200443 Chr.22: 20568487-20568487 on Build GRCh38
rs762376341 Chr.22: 20568478-20568478 on Build GRCh38
rs773874370 Chr.22: 20568485-20568485 on Build GRCh38
rs781079039 Chr.22: 20568535-20568543 on Build GRCh38
rs924234454 Chr.22: 20568609-20568609 on Build GRCh38
rs779974079 Chr.22: 20568532-20568532 on Build GRCh38
rs574249407 Chr.22: 20568573-20568573 on Build GRCh38
rs374565269 Chr.22: 20568643-20568643 on Build GRCh38
rs776925574 Chr.22: 20568620-20568620 on Build GRCh38
rs766656818 Chr.22: 20568500-20568500 on Build GRCh38
rs747546301 Chr.22: 20568671-20568671 on Build GRCh38
rs967021736 Chr.22: 20568550-20568550 on Build GRCh38
rs745827271 Chr.22: 20568549-20568549 on Build GRCh38
rs748960763 Chr.22: 20568568-20568568 on Build GRCh38
rs201038715 Chr.22: 20568639-20568639 on Build GRCh38
rs563844695 Chr.22: 20568480-20568480 on Build GRCh38
rs879233690 Chr.22: 20568525-20568525 on Build GRCh38
rs770175878 Chr.22: 20568682-20568682 on Build GRCh38
rs779384300 Chr.22: 20568526-20568526 on Build GRCh38
rs376642456 Chr.22: 20568505-20568505 on Build GRCh38
rs774065824 Chr.22: 20568476-20568476 on Build GRCh38
rs978480588 Chr.22: 20568552-20568552 on Build GRCh38
rs755210247 Chr.22: 20568516-20568516 on Build GRCh38
rs745974008 Chr.22: 20568537-20568538 on Build GRCh38
rs998522496 Chr.22: 20568511-20568511 on Build GRCh38
rs747940632 Chr.22: 20568633-20568640 on Build GRCh38
rs768410178 Chr.22: 20568568-20568570 on Build GRCh38
rs779729817 Chr.22: 20568565-20568565 on Build GRCh38
rs747609243 Chr.22: 20568587-20568587 on Build GRCh38
rs117948318 Chr.22: 20568611-20568611 on Build GRCh38
rs551506648 Chr.22: 20568653-20568653 on Build GRCh38
rs759775069 Chr.22: 20568622-20568622 on Build GRCh38
rs774411532 Chr.22: 20568567-20568568 on Build GRCh38
rs762520987 Chr.22: 20568472-20568472 on Build GRCh38
rs747803181 Chr.22: 20568659-20568659 on Build GRCh38
rs755874905 Chr.22: 20568553-20568553 on Build GRCh38
rs768460294 Chr.22: 20568571-20568571 on Build GRCh38
rs773788272 Chr.22: 20568602-20568602 on Build GRCh38
rs565016408 Chr.22: 20568672-20568672 on Build GRCh38
rs528653147 Chr.22: 20568642-20568642 on Build GRCh38
rs777034586 Chr.22: 20568673-20568673 on Build GRCh38
rs750087429 Chr.22: 20568460-20568460 on Build GRCh38
rs779897071 Chr.22: 20568645-20568645 on Build GRCh38
rs778664392 Chr.22: 20568658-20568658 on Build GRCh38
rs772654401 Chr.22: 20568670-20568670 on Build GRCh38
rs768158279 Chr.22: 20568474-20568474 on Build GRCh38
rs959116737 Chr.22: 20568563-20568563 on Build GRCh38
rs752815505 Chr.22: 20568518-20568518 on Build GRCh38
rs766751260 Chr.22: 20568614-20568614 on Build GRCh38
rs771649025 Chr.22: 20568664-20568664 on Build GRCh38
rs978540519 Chr.22: 20568539-20568539 on Build GRCh38

View Full Product Details