Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 8891041-8891312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFHAS1
Entrez Gene ID
9258
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
LRRC65, MASL1
UniGene
Hs.379414
Gene Chromosome Location
Chr.8: 8783354-8893621 on Build GRCh38
Transcripts
NM_004225.2 XM_011543852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5714073 c.1961A>C p.N654T Chr.8: 8891098-8891098 on Build GRCh38
5677528 c.1780G>T p.A594S Chr.8: 8891279-8891279 on Build GRCh38
5624687 c.1975C>A p.L659M Chr.8: 8891084-8891084 on Build GRCh38
2789720 c.1897C>T p.R633C Chr.8: 8891162-8891162 on Build GRCh38
282780 c.1910G>A p.R637H Chr.8: 8891149-8891149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367663776 Chr.8: 8891152-8891152 on Build GRCh38
rs772985216 Chr.8: 8891286-8891286 on Build GRCh38
rs765985421 Chr.8: 8891163-8891163 on Build GRCh38
rs146237763 Chr.8: 8891220-8891220 on Build GRCh38
rs768920060 Chr.8: 8891279-8891279 on Build GRCh38
rs772218662 Chr.8: 8891285-8891285 on Build GRCh38
rs930865169 Chr.8: 8891182-8891182 on Build GRCh38
rs538507714 Chr.8: 8891098-8891098 on Build GRCh38
rs150944602 Chr.8: 8891262-8891262 on Build GRCh38
rs759266255 Chr.8: 8891243-8891243 on Build GRCh38
rs757009597 Chr.8: 8891066-8891066 on Build GRCh38
rs867884452 Chr.8: 8891236-8891236 on Build GRCh38
rs892311411 Chr.8: 8891254-8891254 on Build GRCh38
rs889320172 Chr.8: 8891219-8891219 on Build GRCh38
rs1050543235 Chr.8: 8891201-8891201 on Build GRCh38
rs200594818 Chr.8: 8891078-8891078 on Build GRCh38
rs755784449 Chr.8: 8891087-8891087 on Build GRCh38
rs778578585 Chr.8: 8891172-8891172 on Build GRCh38
rs940833007 Chr.8: 8891149-8891149 on Build GRCh38
rs775619271 Chr.8: 8891133-8891133 on Build GRCh38
rs766217777 Chr.8: 8891228-8891228 on Build GRCh38
rs148465761 Chr.8: 8891226-8891226 on Build GRCh38
rs369660366 Chr.8: 8891159-8891159 on Build GRCh38
rs764192422 Chr.8: 8891141-8891141 on Build GRCh38
rs1046066431 Chr.8: 8891118-8891118 on Build GRCh38
rs375316622 Chr.8: 8891248-8891248 on Build GRCh38
rs746268211 Chr.8: 8891196-8891196 on Build GRCh38
rs375503932 Chr.8: 8891282-8891282 on Build GRCh38
rs888776040 Chr.8: 8891210-8891210 on Build GRCh38
rs770574910 Chr.8: 8891292-8891292 on Build GRCh38
rs768668649 Chr.8: 8891214-8891214 on Build GRCh38
rs999279796 Chr.8: 8891245-8891245 on Build GRCh38
rs781412122 Chr.8: 8891060-8891060 on Build GRCh38
rs571544041 Chr.8: 8891127-8891127 on Build GRCh38
rs146307219 Chr.8: 8891084-8891084 on Build GRCh38
rs201593143 Chr.8: 8891106-8891106 on Build GRCh38
rs898230991 Chr.8: 8891153-8891153 on Build GRCh38
rs764834874 Chr.8: 8891244-8891244 on Build GRCh38
rs773026331 Chr.8: 8891223-8891223 on Build GRCh38
rs781608176 Chr.8: 8891193-8891193 on Build GRCh38
rs139956353 Chr.8: 8891093-8891093 on Build GRCh38
rs372323692 Chr.8: 8891281-8891281 on Build GRCh38
rs754487799 Chr.8: 8891168-8891168 on Build GRCh38
rs779599873 Chr.8: 8891091-8891091 on Build GRCh38
rs761755633 Chr.8: 8891148-8891148 on Build GRCh38
rs756390661 Chr.8: 8891263-8891263 on Build GRCh38
rs760236549 Chr.8: 8891189-8891190 on Build GRCh38
rs749538221 Chr.8: 8891208-8891208 on Build GRCh38
rs142559689 Chr.8: 8891249-8891249 on Build GRCh38
rs1533102 Chr.8: 8891207-8891207 on Build GRCh38
rs753573919 Chr.8: 8891165-8891165 on Build GRCh38
rs780399040 Chr.8: 8891272-8891272 on Build GRCh38
rs753481897 Chr.8: 8891242-8891242 on Build GRCh38
rs760668328 Chr.8: 8891287-8891287 on Build GRCh38
rs140782884 Chr.8: 8891275-8891275 on Build GRCh38
rs746228949 Chr.8: 8891218-8891218 on Build GRCh38
rs774226763 Chr.8: 8891143-8891143 on Build GRCh38
rs776625756 Chr.8: 8891293-8891293 on Build GRCh38
rs774423073 Chr.8: 8891216-8891216 on Build GRCh38
rs561174287 Chr.8: 8891094-8891094 on Build GRCh38
rs915781443 Chr.8: 8891103-8891103 on Build GRCh38
rs771362930 Chr.8: 8891109-8891109 on Build GRCh38
rs201372453 Chr.8: 8891190-8891190 on Build GRCh38
rs746099046 Chr.8: 8891120-8891120 on Build GRCh38
rs770178851 Chr.8: 8891251-8891251 on Build GRCh38
rs143510651 Chr.8: 8891139-8891139 on Build GRCh38
rs372091548 Chr.8: 8891154-8891154 on Build GRCh38
rs866682245 Chr.8: 8891209-8891209 on Build GRCh38
rs967374722 Chr.8: 8891234-8891234 on Build GRCh38
rs770029293 Chr.8: 8891198-8891198 on Build GRCh38

View Full Product Details