Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38481627-38481870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIFR
Entrez Gene ID
3977
Gene Name
leukemia inhibitory factor receptor alpha
Gene Aliases
CD118, LIF-R, SJS2, STWS, SWS
UniGene
Hs.133421
Gene Chromosome Location
Chr.5: 38474963-38606290 on Build GRCh38
Transcripts
NM_001127671.1 NM_002310.5 XM_011514040.2 XM_011514042.2 XM_017009462.1 XM_017009463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3920020 c.3194C>T p.S1065F Chr.5: 38481695-38481695 on Build GRCh38
33086 c.3204T>G p.F1068L Chr.5: 38481685-38481685 on Build GRCh38
5720320 c.3251G>A p.G1084E Chr.5: 38481638-38481638 on Build GRCh38
4487363 c.3179C>T p.P1060L Chr.5: 38481710-38481710 on Build GRCh38
1437432 c.3086G>A p.R1029K Chr.5: 38481803-38481803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772164575 Chr.5: 38481794-38481794 on Build GRCh38
rs548778843 Chr.5: 38481828-38481828 on Build GRCh38
rs748122146 Chr.5: 38481790-38481790 on Build GRCh38
rs886060626 Chr.5: 38481807-38481807 on Build GRCh38
rs762094729 Chr.5: 38481709-38481709 on Build GRCh38
rs867316235 Chr.5: 38481666-38481666 on Build GRCh38
rs138418444 Chr.5: 38481802-38481802 on Build GRCh38
rs373729193 Chr.5: 38481712-38481712 on Build GRCh38
rs201226332 Chr.5: 38481722-38481722 on Build GRCh38
rs765137025 Chr.5: 38481767-38481767 on Build GRCh38
rs147058538 Chr.5: 38481668-38481668 on Build GRCh38
rs776513402 Chr.5: 38481657-38481658 on Build GRCh38
rs943207929 Chr.5: 38481650-38481650 on Build GRCh38
rs766709490 Chr.5: 38481805-38481805 on Build GRCh38
rs1012355482 Chr.5: 38481837-38481837 on Build GRCh38
rs751710397 Chr.5: 38481831-38481831 on Build GRCh38
rs377647437 Chr.5: 38481723-38481723 on Build GRCh38
rs756805690 Chr.5: 38481754-38481754 on Build GRCh38
rs768761408 Chr.5: 38481672-38481672 on Build GRCh38
rs768547048 Chr.5: 38481781-38481781 on Build GRCh38
rs780926951 Chr.5: 38481756-38481756 on Build GRCh38
rs776919303 Chr.5: 38481810-38481810 on Build GRCh38
rs745548189 Chr.5: 38481757-38481757 on Build GRCh38
rs537706381 Chr.5: 38481829-38481829 on Build GRCh38
rs763655688 Chr.5: 38481746-38481746 on Build GRCh38
rs755877357 Chr.5: 38481834-38481834 on Build GRCh38
rs765596913 Chr.5: 38481812-38481812 on Build GRCh38
rs774354232 Chr.5: 38481787-38481787 on Build GRCh38
rs773424045 Chr.5: 38481718-38481718 on Build GRCh38
rs368698516 Chr.5: 38481692-38481692 on Build GRCh38
rs749221244 Chr.5: 38481780-38481780 on Build GRCh38
rs755926450 Chr.5: 38481758-38481758 on Build GRCh38
rs1003123167 Chr.5: 38481762-38481762 on Build GRCh38
rs755477526 Chr.5: 38481742-38481742 on Build GRCh38
rs370859972 Chr.5: 38481741-38481741 on Build GRCh38
rs760083976 Chr.5: 38481811-38481811 on Build GRCh38
rs1035689314 Chr.5: 38481795-38481795 on Build GRCh38
rs753260599 Chr.5: 38481819-38481819 on Build GRCh38
rs773385403 Chr.5: 38481797-38481797 on Build GRCh38
rs1012911330 Chr.5: 38481803-38481803 on Build GRCh38
rs766750299 Chr.5: 38481734-38481734 on Build GRCh38
rs751088321 Chr.5: 38481748-38481748 on Build GRCh38

View Full Product Details