Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1895365-1895600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRPA
Entrez Gene ID
140885
Gene Name
signal regulatory protein alpha
Gene Aliases
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP
UniGene
Hs.581021
Gene Chromosome Location
Chr.20: 1894117-1939895 on Build GRCh38
Transcripts
NM_001040022.1 NM_001040023.1 NM_080792.2 XM_005260669.3 XM_005260670.3 XM_006723545.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319689 c.45C>T p.L15L Chr.20: 1895492-1895492 on Build GRCh38
5449399 c.58G>A p.A20T Chr.20: 1895505-1895505 on Build GRCh38
5513600 c.79+8delC p.? Chr.20: 1895534-1895534 on Build GRCh38
4512343 c.8C>T p.P3L Chr.20: 1895455-1895455 on Build GRCh38
6410093 c.37C>A p.P13T Chr.20: 1895484-1895484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989697705 Chr.20: 1895539-1895539 on Build GRCh38
rs755781590 Chr.20: 1895467-1895467 on Build GRCh38
rs750995601 Chr.20: 1895433-1895433 on Build GRCh38
rs931948826 Chr.20: 1895540-1895540 on Build GRCh38
rs981548852 Chr.20: 1895398-1895398 on Build GRCh38
rs1025165674 Chr.20: 1895443-1895443 on Build GRCh38
rs967304709 Chr.20: 1895557-1895557 on Build GRCh38
rs757862537 Chr.20: 1895486-1895486 on Build GRCh38
rs562529545 Chr.20: 1895567-1895567 on Build GRCh38
rs1033320320 Chr.20: 1895492-1895492 on Build GRCh38
rs968099700 Chr.20: 1895462-1895462 on Build GRCh38
rs941914854 Chr.20: 1895389-1895389 on Build GRCh38
rs923654765 Chr.20: 1895498-1895498 on Build GRCh38
rs990899951 Chr.20: 1895427-1895427 on Build GRCh38
rs112674043 Chr.20: 1895439-1895439 on Build GRCh38
rs1001882194 Chr.20: 1895470-1895470 on Build GRCh38
rs759597616 Chr.20: 1895533-1895534 on Build GRCh38
rs958947685 Chr.20: 1895426-1895426 on Build GRCh38
rs1040190802 Chr.20: 1895516-1895516 on Build GRCh38
rs962215784 Chr.20: 1895397-1895397 on Build GRCh38
rs1037506845 Chr.20: 1895395-1895395 on Build GRCh38
rs887810198 Chr.20: 1895582-1895582 on Build GRCh38
rs922692567 Chr.20: 1895575-1895575 on Build GRCh38
rs916282843 Chr.20: 1895440-1895440 on Build GRCh38
rs976934372 Chr.20: 1895566-1895566 on Build GRCh38
rs935169449 Chr.20: 1895507-1895507 on Build GRCh38
rs1036372220 Chr.20: 1895403-1895403 on Build GRCh38
rs781759027 Chr.20: 1895548-1895548 on Build GRCh38
rs923187821 Chr.20: 1895525-1895525 on Build GRCh38

View Full Product Details