Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42437195-42437438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC20A2
Entrez Gene ID
6575
Gene Name
solute carrier family 20 member 2
Gene Aliases
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
UniGene
Hs.653173
Gene Chromosome Location
Chr.8: 42416462-42542213 on Build GRCh38
Transcripts
NM_001257180.1 NM_001257181.1 NM_006749.4 XM_005273613.3 XM_005273615.3 XM_006716390.3 XM_006716391.3 XM_017013748.1 XM_017013749.1 XM_017013750.1 XM_017013751.1 XM_017013752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874720 c.1223C>T p.S408L Chr.8: 42437289-42437289 on Build GRCh38
750319 c.1102G>T p.E368* Chr.8: 42437410-42437410 on Build GRCh38
3382257 c.1285C>T p.R429C Chr.8: 42437227-42437227 on Build GRCh38
1457170 c.1214C>T p.A405V Chr.8: 42437298-42437298 on Build GRCh38
6516138 c.1149C>T p.N383N Chr.8: 42437363-42437363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377708700 Chr.8: 42437336-42437336 on Build GRCh38
rs115234947 Chr.8: 42437258-42437258 on Build GRCh38
rs116194858 Chr.8: 42437314-42437314 on Build GRCh38
rs756419424 Chr.8: 42437324-42437324 on Build GRCh38
rs147905358 Chr.8: 42437402-42437402 on Build GRCh38
rs527716146 Chr.8: 42437389-42437389 on Build GRCh38
rs767994782 Chr.8: 42437315-42437315 on Build GRCh38
rs920439006 Chr.8: 42437224-42437224 on Build GRCh38
rs201993727 Chr.8: 42437297-42437297 on Build GRCh38
rs775204334 Chr.8: 42437359-42437360 on Build GRCh38
rs757370573 Chr.8: 42437353-42437353 on Build GRCh38
rs1055093172 Chr.8: 42437379-42437379 on Build GRCh38
rs751972151 Chr.8: 42437226-42437226 on Build GRCh38
rs1013714218 Chr.8: 42437414-42437414 on Build GRCh38
rs746862912 Chr.8: 42437253-42437253 on Build GRCh38
rs574609540 Chr.8: 42437252-42437252 on Build GRCh38
rs781183228 Chr.8: 42437395-42437395 on Build GRCh38
rs199935692 Chr.8: 42437411-42437411 on Build GRCh38
rs772953742 Chr.8: 42437301-42437301 on Build GRCh38
rs114636687 Chr.8: 42437378-42437378 on Build GRCh38
rs140966410 Chr.8: 42437375-42437375 on Build GRCh38
rs747841902 Chr.8: 42437289-42437289 on Build GRCh38
rs148397659 Chr.8: 42437231-42437231 on Build GRCh38
rs753034307 Chr.8: 42437317-42437317 on Build GRCh38
rs943755261 Chr.8: 42437282-42437282 on Build GRCh38
rs757283528 Chr.8: 42437251-42437251 on Build GRCh38
rs762584113 Chr.8: 42437305-42437305 on Build GRCh38
rs917367482 Chr.8: 42437290-42437290 on Build GRCh38
rs199804961 Chr.8: 42437230-42437230 on Build GRCh38
rs754838969 Chr.8: 42437391-42437391 on Build GRCh38
rs773208227 Chr.8: 42437227-42437227 on Build GRCh38
rs756327679 Chr.8: 42437233-42437233 on Build GRCh38
rs768552976 Chr.8: 42437257-42437257 on Build GRCh38
rs749389443 Chr.8: 42437249-42437249 on Build GRCh38
rs532263007 Chr.8: 42437313-42437313 on Build GRCh38
rs766931477 Chr.8: 42437222-42437222 on Build GRCh38
rs777876972 Chr.8: 42437236-42437236 on Build GRCh38
rs972449015 Chr.8: 42437287-42437287 on Build GRCh38
rs200010919 Chr.8: 42437367-42437367 on Build GRCh38
rs769623826 Chr.8: 42437354-42437355 on Build GRCh38

View Full Product Details