Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25673050-25673281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC34A2
Entrez Gene ID
10568
Gene Name
solute carrier family 34 member 2
Gene Aliases
NAPI-3B, NAPI-IIb, NPTIIb
UniGene
Hs.479372
Gene Chromosome Location
Chr.4: 25655813-25678748 on Build GRCh38
Transcripts
NM_001177998.1 NM_001177999.1 NM_006424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167273 c.1107C>G p.L369L Chr.4: 25673145-25673145 on Build GRCh38
315304 c.1122G>T p.L374L Chr.4: 25673160-25673160 on Build GRCh38
5401054 c.1173C>T p.L391L Chr.4: 25673211-25673211 on Build GRCh38
6100140 c.1205C>A p.T402N Chr.4: 25673243-25673243 on Build GRCh38
3603099 c.1113C>T p.L371L Chr.4: 25673151-25673151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768442620 Chr.4: 25673137-25673137 on Build GRCh38
rs760439582 Chr.4: 25673073-25673073 on Build GRCh38
rs34723230 Chr.4: 25673127-25673127 on Build GRCh38
rs765673564 Chr.4: 25673230-25673230 on Build GRCh38
rs1019797044 Chr.4: 25673139-25673139 on Build GRCh38
rs138755203 Chr.4: 25673117-25673117 on Build GRCh38
rs546457472 Chr.4: 25673118-25673118 on Build GRCh38
rs763519879 Chr.4: 25673247-25673247 on Build GRCh38
rs752707048 Chr.4: 25673115-25673115 on Build GRCh38
rs868371878 Chr.4: 25673204-25673204 on Build GRCh38
rs771701778 Chr.4: 25673186-25673186 on Build GRCh38
rs757372305 Chr.4: 25673263-25673263 on Build GRCh38
rs945164408 Chr.4: 25673084-25673084 on Build GRCh38
rs748558105 Chr.4: 25673213-25673214 on Build GRCh38
rs765763683 Chr.4: 25673109-25673109 on Build GRCh38
rs967172413 Chr.4: 25673076-25673076 on Build GRCh38
rs758484073 Chr.4: 25673116-25673116 on Build GRCh38
rs377419850 Chr.4: 25673091-25673091 on Build GRCh38
rs141905062 Chr.4: 25673245-25673245 on Build GRCh38
rs148080437 Chr.4: 25673096-25673096 on Build GRCh38
rs770634792 Chr.4: 25673212-25673212 on Build GRCh38
rs917641107 Chr.4: 25673080-25673081 on Build GRCh38
rs747607173 Chr.4: 25673157-25673157 on Build GRCh38
rs370229349 Chr.4: 25673255-25673255 on Build GRCh38
rs764129224 Chr.4: 25673254-25673254 on Build GRCh38
rs772886127 Chr.4: 25673207-25673207 on Build GRCh38
rs541971815 Chr.4: 25673223-25673223 on Build GRCh38
rs576225512 Chr.4: 25673140-25673140 on Build GRCh38
rs560657112 Chr.4: 25673224-25673224 on Build GRCh38
rs760218005 Chr.4: 25673209-25673209 on Build GRCh38
rs759907304 Chr.4: 25673102-25673102 on Build GRCh38
rs749131631 Chr.4: 25673130-25673130 on Build GRCh38
rs915538683 Chr.4: 25673146-25673146 on Build GRCh38

View Full Product Details