Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124249598-124249871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8G1
Entrez Gene ID
26494
Gene Name
olfactory receptor family 8 subfamily G member 1 (gene/pseudogene)
Gene Aliases
HSTPCR25, OR8G1P, TPCR25
UniGene
Hs.448172
Gene Chromosome Location
Chr.11: 124249676-124250611 on Build GRCh38
Transcripts
NM_001002905.1 NR_045681.1 XM_011542662.2 XM_011542663.2 XM_011542664.2 XM_011542665.2 XM_011542666.2 XM_011542667.2

Gene Symbol
OR8G5
Entrez Gene ID
219865
Gene Name
olfactory receptor family 8 subfamily G member 5
Gene Aliases
OR11-298, OR8G5P, OR8G6
UniGene
Hs.722725
Gene Chromosome Location
Chr.11: 124241158-124323283 on Build GRCh38
Transcripts
NM_001002905.1 NR_045681.1 XM_011542662.2 XM_011542663.2 XM_011542664.2 XM_011542665.2 XM_011542666.2 XM_011542667.2

SNP Detail

SNP ID Chromosome Location
rs969257286 Chr.11: 124249628-124249628 on Build GRCh38
rs766977639 Chr.11: 124249644-124249644 on Build GRCh38
rs759085261 Chr.11: 124249668-124249668 on Build GRCh38
rs756992232 Chr.11: 124249639-124249641 on Build GRCh38
rs778054627 Chr.11: 124249835-124249835 on Build GRCh38
rs757103806 Chr.11: 124249699-124249699 on Build GRCh38
rs750403205 Chr.11: 124249718-124249718 on Build GRCh38
rs186498061 Chr.11: 124249669-124249669 on Build GRCh38
rs1045341057 Chr.11: 124249647-124249647 on Build GRCh38
rs760195365 Chr.11: 124249781-124249781 on Build GRCh38
rs748248536 Chr.11: 124249730-124249730 on Build GRCh38
rs995669156 Chr.11: 124249726-124249726 on Build GRCh38
rs960576508 Chr.11: 124249670-124249670 on Build GRCh38
rs528413850 Chr.11: 124249823-124249823 on Build GRCh38
rs752802783 Chr.11: 124249825-124249825 on Build GRCh38
rs781358946 Chr.11: 124249722-124249722 on Build GRCh38
rs138496920 Chr.11: 124249813-124249813 on Build GRCh38
rs1054637892 Chr.11: 124249630-124249630 on Build GRCh38
rs376965349 Chr.11: 124249641-124249641 on Build GRCh38
rs370541814 Chr.11: 124249745-124249745 on Build GRCh38
rs760133703 Chr.11: 124249772-124249772 on Build GRCh38
rs771491415 Chr.11: 124249752-124249752 on Build GRCh38
rs753681705 Chr.11: 124249695-124249695 on Build GRCh38
rs879712897 Chr.11: 124249704-124249705 on Build GRCh38
rs765019603 Chr.11: 124249811-124249811 on Build GRCh38
rs765072062 Chr.11: 124249802-124249802 on Build GRCh38
rs201813599 Chr.11: 124249784-124249784 on Build GRCh38
rs542660975 Chr.11: 124249812-124249812 on Build GRCh38
rs977771623 Chr.11: 124249837-124249837 on Build GRCh38
rs949831599 Chr.11: 124249826-124249826 on Build GRCh38
rs181862385 Chr.11: 124249626-124249626 on Build GRCh38
rs577844540 Chr.11: 124249685-124249685 on Build GRCh38
rs775063655 Chr.11: 124249655-124249655 on Build GRCh38
rs760406916 Chr.11: 124249677-124249677 on Build GRCh38
rs1004912387 Chr.11: 124249679-124249679 on Build GRCh38
rs545481117 Chr.11: 124249737-124249737 on Build GRCh38
rs973722002 Chr.11: 124249790-124249790 on Build GRCh38
rs575554716 Chr.11: 124249807-124249807 on Build GRCh38
rs755003193 Chr.11: 124249843-124249843 on Build GRCh38
rs777912617 Chr.11: 124249750-124249750 on Build GRCh38
rs752242239 Chr.11: 124249672-124249672 on Build GRCh38
rs368654832 Chr.11: 124249795-124249795 on Build GRCh38
rs778820261 Chr.11: 124249702-124249702 on Build GRCh38
rs771640961 Chr.11: 124249774-124249774 on Build GRCh38
rs147084212 Chr.11: 124249779-124249779 on Build GRCh38
rs756190344 Chr.11: 124249832-124249832 on Build GRCh38
rs199680811 Chr.11: 124249753-124249753 on Build GRCh38
rs748764124 Chr.11: 124249629-124249632 on Build GRCh38
rs749580936 Chr.11: 124249846-124249846 on Build GRCh38
rs932867495 Chr.11: 124249808-124249808 on Build GRCh38
rs774986408 Chr.11: 124249762-124249762 on Build GRCh38

View Full Product Details