Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40432494-40432767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM47
Entrez Gene ID
54502
Gene Name
RNA binding motif protein 47
Gene Aliases
NET18
UniGene
Hs.518727
Gene Chromosome Location
Chr.4: 40423255-40630911 on Build GRCh38
Transcripts
NM_001098634.1 NM_019027.3 XM_005248103.3 XM_005248107.3 XM_005248108.3 XM_005248109.4 XM_011513704.2 XM_011513707.2 XM_011513708.2 XM_017008304.1 XM_017008305.1 XM_017008306.1 XM_017008307.1 XM_017008308.1 XM_017008309.1 XM_017008310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4720748 c.1278_1289del12 p.A430_A433delAAAA Chr.4: 40432697-40432708 on Build GRCh38
4720745 c.1290_1298delCGCAGCCGC p.A431_A433delAAA Chr.4: 40432688-40432696 on Build GRCh38
163955 c.1270G>A p.A424T Chr.4: 40432716-40432716 on Build GRCh38
4332679 c.1269C>T p.A423A Chr.4: 40432717-40432717 on Build GRCh38
4332652 c.1295C>T p.A432V Chr.4: 40432691-40432691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765476683 Chr.4: 40432702-40432703 on Build GRCh38
rs762999861 Chr.4: 40432632-40432632 on Build GRCh38
rs774944998 Chr.4: 40432690-40432690 on Build GRCh38
rs763890818 Chr.4: 40432736-40432736 on Build GRCh38
rs971755083 Chr.4: 40432705-40432705 on Build GRCh38
rs762733129 Chr.4: 40432687-40432688 on Build GRCh38
rs567159198 Chr.4: 40432720-40432720 on Build GRCh38
rs567926183 Chr.4: 40432550-40432550 on Build GRCh38
rs767795669 Chr.4: 40432666-40432666 on Build GRCh38
rs759043120 Chr.4: 40432696-40432697 on Build GRCh38
rs41265677 Chr.4: 40432608-40432608 on Build GRCh38
rs200151630 Chr.4: 40432630-40432630 on Build GRCh38
rs768665539 Chr.4: 40432711-40432711 on Build GRCh38
rs530922627 Chr.4: 40432542-40432542 on Build GRCh38
rs528269773 Chr.4: 40432697-40432708 on Build GRCh38
rs997398374 Chr.4: 40432534-40432534 on Build GRCh38
rs751753386 Chr.4: 40432730-40432730 on Build GRCh38
rs3752824 Chr.4: 40432717-40432717 on Build GRCh38
rs1039290692 Chr.4: 40432521-40432521 on Build GRCh38
rs752324664 Chr.4: 40432700-40432708 on Build GRCh38
rs780593044 Chr.4: 40432720-40432721 on Build GRCh38
rs370564777 Chr.4: 40432697-40432697 on Build GRCh38
rs756689460 Chr.4: 40432741-40432741 on Build GRCh38
rs750942039 Chr.4: 40432706-40432708 on Build GRCh38
rs967393710 Chr.4: 40432646-40432646 on Build GRCh38
rs748009887 Chr.4: 40432686-40432694 on Build GRCh38
rs781516629 Chr.4: 40432624-40432624 on Build GRCh38
rs762758618 Chr.4: 40432696-40432696 on Build GRCh38
rs756926169 Chr.4: 40432694-40432694 on Build GRCh38
rs755929230 Chr.4: 40432708-40432708 on Build GRCh38
rs1004571061 Chr.4: 40432546-40432546 on Build GRCh38
rs749487596 Chr.4: 40432598-40432598 on Build GRCh38
rs758772454 Chr.4: 40432709-40432720 on Build GRCh38
rs757863574 Chr.4: 40432607-40432607 on Build GRCh38
rs768777352 Chr.4: 40432645-40432645 on Build GRCh38
rs756413413 Chr.4: 40432698-40432698 on Build GRCh38
rs200374378 Chr.4: 40432689-40432689 on Build GRCh38
rs746183897 Chr.4: 40432625-40432625 on Build GRCh38
rs928769504 Chr.4: 40432552-40432552 on Build GRCh38
rs775331815 Chr.4: 40432725-40432725 on Build GRCh38
rs751114219 Chr.4: 40432739-40432739 on Build GRCh38
rs780785396 Chr.4: 40432695-40432695 on Build GRCh38
rs146911971 Chr.4: 40432667-40432667 on Build GRCh38
rs974570838 Chr.4: 40432693-40432693 on Build GRCh38
rs436166 Chr.4: 40432723-40432723 on Build GRCh38
rs779245534 Chr.4: 40432652-40432652 on Build GRCh38
rs759333854 Chr.4: 40432669-40432669 on Build GRCh38
rs1051371940 Chr.4: 40432539-40432539 on Build GRCh38
rs781538042 Chr.4: 40432685-40432693 on Build GRCh38
rs769490809 Chr.4: 40432688-40432696 on Build GRCh38
rs570958133 Chr.4: 40432611-40432611 on Build GRCh38
rs774510869 Chr.4: 40432714-40432714 on Build GRCh38
rs926029576 Chr.4: 40432687-40432687 on Build GRCh38
rs767171365 Chr.4: 40432748-40432748 on Build GRCh38
rs938475554 Chr.4: 40432709-40432709 on Build GRCh38
rs762185282 Chr.4: 40432703-40432723 on Build GRCh38
rs1001686433 Chr.4: 40432585-40432585 on Build GRCh38
rs776185295 Chr.4: 40432523-40432523 on Build GRCh38
rs547575066 Chr.4: 40432699-40432707 on Build GRCh38
rs762072583 Chr.4: 40432664-40432664 on Build GRCh38
rs780492136 Chr.4: 40432704-40432704 on Build GRCh38
rs773506304 Chr.4: 40432719-40432719 on Build GRCh38
rs748134090 Chr.4: 40432716-40432716 on Build GRCh38
rs1034933514 Chr.4: 40432595-40432595 on Build GRCh38
rs770316901 Chr.4: 40432626-40432626 on Build GRCh38
rs564837143 Chr.4: 40432688-40432708 on Build GRCh38
rs940407838 Chr.4: 40432548-40432548 on Build GRCh38
rs376656036 Chr.4: 40432638-40432638 on Build GRCh38
rs777351744 Chr.4: 40432610-40432610 on Build GRCh38
rs898923536 Chr.4: 40432551-40432551 on Build GRCh38
rs771328457 Chr.4: 40432614-40432614 on Build GRCh38
rs201549023 Chr.4: 40432685-40432685 on Build GRCh38
rs749771006 Chr.4: 40432710-40432710 on Build GRCh38

View Full Product Details