Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105177521-105177776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT14
Entrez Gene ID
256281
Gene Name
nudix hydrolase 14
Gene Aliases
UGPP, UGPPase
UniGene
Hs.526432
Gene Chromosome Location
Chr.14: 105172937-105181323 on Build GRCh38
Transcripts
NM_001318380.1 NM_177533.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
953962 c.116C>T p.T39M Chr.14: 105177701-105177701 on Build GRCh38
5927715 c.116C>G p.T39R Chr.14: 105177701-105177701 on Build GRCh38
416264 c.82-1G>A p.? Chr.14: 105177736-105177736 on Build GRCh38
5745245 c.118C>T p.H40Y Chr.14: 105177699-105177699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374593034 Chr.14: 105177691-105177691 on Build GRCh38
rs752515273 Chr.14: 105177709-105177709 on Build GRCh38
rs587607998 Chr.14: 105177590-105177590 on Build GRCh38
rs769372298 Chr.14: 105177743-105177743 on Build GRCh38
rs761502172 Chr.14: 105177687-105177687 on Build GRCh38
rs1025166046 Chr.14: 105177597-105177597 on Build GRCh38
rs776639089 Chr.14: 105177699-105177699 on Build GRCh38
rs777285020 Chr.14: 105177689-105177689 on Build GRCh38
rs117495827 Chr.14: 105177755-105177755 on Build GRCh38
rs769951564 Chr.14: 105177650-105177650 on Build GRCh38
rs10139839 Chr.14: 105177639-105177639 on Build GRCh38
rs935994223 Chr.14: 105177581-105177581 on Build GRCh38
rs780631746 Chr.14: 105177670-105177670 on Build GRCh38
rs587657786 Chr.14: 105177700-105177700 on Build GRCh38
rs749872659 Chr.14: 105177751-105177751 on Build GRCh38
rs587609526 Chr.14: 105177641-105177641 on Build GRCh38
rs891273767 Chr.14: 105177671-105177671 on Build GRCh38
rs994473441 Chr.14: 105177736-105177736 on Build GRCh38
rs990198235 Chr.14: 105177601-105177601 on Build GRCh38
rs749698896 Chr.14: 105177683-105177683 on Build GRCh38
rs977353440 Chr.14: 105177552-105177552 on Build GRCh38
rs1027069701 Chr.14: 105177674-105177674 on Build GRCh38
rs774720226 Chr.14: 105177718-105177718 on Build GRCh38
rs150509138 Chr.14: 105177721-105177721 on Build GRCh38
rs760202132 Chr.14: 105177694-105177694 on Build GRCh38
rs765751024 Chr.14: 105177698-105177698 on Build GRCh38
rs757590724 Chr.14: 105177710-105177710 on Build GRCh38
rs773094675 Chr.14: 105177754-105177754 on Build GRCh38
rs921316053 Chr.14: 105177616-105177616 on Build GRCh38
rs924620360 Chr.14: 105177577-105177577 on Build GRCh38
rs1034353101 Chr.14: 105177551-105177551 on Build GRCh38
rs750622332 Chr.14: 105177739-105177739 on Build GRCh38
rs747638127 Chr.14: 105177582-105177582 on Build GRCh38
rs769069619 Chr.14: 105177685-105177685 on Build GRCh38
rs587720639 Chr.14: 105177640-105177640 on Build GRCh38
rs776179149 Chr.14: 105177701-105177701 on Build GRCh38

View Full Product Details