Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85081199-85081436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR63
Entrez Gene ID
126820
Gene Name
WD repeat domain 63
Gene Aliases
DIC3, NYD-SP29
UniGene
Hs.97933
Gene Chromosome Location
Chr.1: 85062298-85133138 on Build GRCh38
Transcripts
NM_001288563.1 NM_145172.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444281 c.278T>A p.I93N Chr.1: 85081408-85081408 on Build GRCh38
912643 c.195T>C p.P65P Chr.1: 85081325-85081325 on Build GRCh38
1344575 c.163C>T p.R55* Chr.1: 85081293-85081293 on Build GRCh38
4447474 c.285+3A>G p.? Chr.1: 85081418-85081418 on Build GRCh38
4530425 c.169G>A p.D57N Chr.1: 85081299-85081299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780969830 Chr.1: 85081374-85081374 on Build GRCh38
rs140825967 Chr.1: 85081339-85081339 on Build GRCh38
rs752700346 Chr.1: 85081351-85081351 on Build GRCh38
rs770637189 Chr.1: 85081284-85081284 on Build GRCh38
rs757028946 Chr.1: 85081368-85081368 on Build GRCh38
rs760779989 Chr.1: 85081344-85081344 on Build GRCh38
rs767717241 Chr.1: 85081336-85081336 on Build GRCh38
rs752361338 Chr.1: 85081350-85081350 on Build GRCh38
rs575950333 Chr.1: 85081365-85081365 on Build GRCh38
rs780916665 Chr.1: 85081288-85081288 on Build GRCh38
rs12088304 Chr.1: 85081367-85081367 on Build GRCh38
rs181501140 Chr.1: 85081278-85081278 on Build GRCh38
rs113771253 Chr.1: 85081330-85081330 on Build GRCh38
rs746669361 Chr.1: 85081273-85081273 on Build GRCh38
rs145459682 Chr.1: 85081293-85081293 on Build GRCh38
rs755679606 Chr.1: 85081397-85081397 on Build GRCh38
rs189167401 Chr.1: 85081244-85081244 on Build GRCh38
rs745408608 Chr.1: 85081395-85081395 on Build GRCh38
rs374125994 Chr.1: 85081366-85081366 on Build GRCh38
rs763321300 Chr.1: 85081320-85081320 on Build GRCh38
rs201744091 Chr.1: 85081301-85081301 on Build GRCh38
rs370422920 Chr.1: 85081294-85081294 on Build GRCh38
rs762089342 Chr.1: 85081334-85081334 on Build GRCh38
rs774632932 Chr.1: 85081324-85081324 on Build GRCh38
rs752477436 Chr.1: 85081234-85081234 on Build GRCh38
rs777659842 Chr.1: 85081257-85081257 on Build GRCh38
rs776785603 Chr.1: 85081323-85081323 on Build GRCh38
rs767139961 Chr.1: 85081270-85081270 on Build GRCh38

View Full Product Details