Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60927307-60927576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132A
Entrez Gene ID
54972
Gene Name
transmembrane protein 132A
Gene Aliases
GBP, HSPA5BP1
UniGene
Hs.118552
Gene Chromosome Location
Chr.11: 60924441-60937159 on Build GRCh38
Transcripts
NM_017870.3 NM_178031.2 XM_017017951.1 XM_017017952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929330 c.302T>C p.F101S Chr.11: 60927405-60927405 on Build GRCh38
7093250 c.230G>A p.R77Q Chr.11: 60927333-60927333 on Build GRCh38
6853154 c.283C>T p.R95W Chr.11: 60927386-60927386 on Build GRCh38
1604842 c.284G>C p.R95P Chr.11: 60927387-60927387 on Build GRCh38
3368453 c.240C>A p.T80T Chr.11: 60927343-60927343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052895927 Chr.11: 60927477-60927477 on Build GRCh38
rs1033219557 Chr.11: 60927494-60927494 on Build GRCh38
rs774731947 Chr.11: 60927552-60927552 on Build GRCh38
rs557676655 Chr.11: 60927427-60927427 on Build GRCh38
rs750127054 Chr.11: 60927466-60927466 on Build GRCh38
rs764016345 Chr.11: 60927327-60927327 on Build GRCh38
rs976230053 Chr.11: 60927356-60927356 on Build GRCh38
rs375305600 Chr.11: 60927434-60927434 on Build GRCh38
rs753559216 Chr.11: 60927333-60927333 on Build GRCh38
rs369141015 Chr.11: 60927452-60927452 on Build GRCh38
rs373433085 Chr.11: 60927430-60927430 on Build GRCh38
rs376128600 Chr.11: 60927464-60927464 on Build GRCh38
rs199994281 Chr.11: 60927435-60927435 on Build GRCh38
rs200168099 Chr.11: 60927393-60927393 on Build GRCh38
rs61739595 Chr.11: 60927349-60927349 on Build GRCh38
rs1034671267 Chr.11: 60927359-60927359 on Build GRCh38
rs1006965411 Chr.11: 60927544-60927544 on Build GRCh38
rs200931303 Chr.11: 60927346-60927346 on Build GRCh38
rs773906622 Chr.11: 60927386-60927386 on Build GRCh38
rs953593206 Chr.11: 60927517-60927517 on Build GRCh38
rs777273092 Chr.11: 60927330-60927330 on Build GRCh38
rs373140123 Chr.11: 60927463-60927463 on Build GRCh38
rs1009287748 Chr.11: 60927387-60927387 on Build GRCh38
rs1011327385 Chr.11: 60927497-60927497 on Build GRCh38
rs554305324 Chr.11: 60927382-60927382 on Build GRCh38
rs371122841 Chr.11: 60927398-60927398 on Build GRCh38
rs761475835 Chr.11: 60927462-60927462 on Build GRCh38
rs746591873 Chr.11: 60927358-60927363 on Build GRCh38
rs778263099 Chr.11: 60927377-60927377 on Build GRCh38
rs571144274 Chr.11: 60927493-60927493 on Build GRCh38
rs953771287 Chr.11: 60927508-60927508 on Build GRCh38
rs760720801 Chr.11: 60927445-60927445 on Build GRCh38
rs763880474 Chr.11: 60927446-60927446 on Build GRCh38
rs967508291 Chr.11: 60927424-60927424 on Build GRCh38
rs76466797 Chr.11: 60927371-60927371 on Build GRCh38
rs756578318 Chr.11: 60927372-60927372 on Build GRCh38
rs1021759206 Chr.11: 60927507-60927507 on Build GRCh38
rs149428442 Chr.11: 60927353-60927353 on Build GRCh38
rs757855854 Chr.11: 60927362-60927362 on Build GRCh38

View Full Product Details