Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13109484-13109719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT1
Entrez Gene ID
55621
Gene Name
tRNA methyltransferase 1
Gene Aliases
TRM1
UniGene
Hs.515169
Gene Chromosome Location
Chr.19: 13104900-13117568 on Build GRCh38
Transcripts
NM_001136035.2 NM_001142554.1 NM_017722.3 XM_006722793.2 XM_011528124.1 XM_011528125.1 XM_017026944.1 XM_017026945.1 XM_017026946.1 XM_017026947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205131 c.1303A>G p.I435V Chr.19: 13109558-13109558 on Build GRCh38
4400108 c.1220T>A p.F407Y Chr.19: 13109641-13109641 on Build GRCh38
1230464 c.1247G>T p.S416I Chr.19: 13109614-13109614 on Build GRCh38
6808932 c.1177-8G>A p.? Chr.19: 13109692-13109692 on Build GRCh38
7208291 c.1185delC p.M397fs*19 Chr.19: 13109676-13109676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756464419 Chr.19: 13109514-13109514 on Build GRCh38
rs370969066 Chr.19: 13109505-13109505 on Build GRCh38
rs369032998 Chr.19: 13109698-13109698 on Build GRCh38
rs368153051 Chr.19: 13109576-13109576 on Build GRCh38
rs972097341 Chr.19: 13109622-13109622 on Build GRCh38
rs143782952 Chr.19: 13109632-13109632 on Build GRCh38
rs367719709 Chr.19: 13109691-13109691 on Build GRCh38
rs762917024 Chr.19: 13109550-13109550 on Build GRCh38
rs201235025 Chr.19: 13109671-13109671 on Build GRCh38
rs200130273 Chr.19: 13109541-13109541 on Build GRCh38
rs770558782 Chr.19: 13109531-13109531 on Build GRCh38
rs749769402 Chr.19: 13109524-13109524 on Build GRCh38
rs200813203 Chr.19: 13109530-13109530 on Build GRCh38
rs759329844 Chr.19: 13109539-13109539 on Build GRCh38
rs77560275 Chr.19: 13109692-13109692 on Build GRCh38
rs375036527 Chr.19: 13109660-13109660 on Build GRCh38
rs748001131 Chr.19: 13109693-13109693 on Build GRCh38
rs919222213 Chr.19: 13109624-13109624 on Build GRCh38
rs761813897 Chr.19: 13109562-13109562 on Build GRCh38
rs750731049 Chr.19: 13109512-13109512 on Build GRCh38
rs147243880 Chr.19: 13109633-13109633 on Build GRCh38
rs776425346 Chr.19: 13109538-13109538 on Build GRCh38
rs147474165 Chr.19: 13109616-13109616 on Build GRCh38
rs138303657 Chr.19: 13109618-13109618 on Build GRCh38
rs754172388 Chr.19: 13109636-13109636 on Build GRCh38
rs751613445 Chr.19: 13109561-13109561 on Build GRCh38
rs138022726 Chr.19: 13109568-13109568 on Build GRCh38
rs780362420 Chr.19: 13109575-13109575 on Build GRCh38
rs755300048 Chr.19: 13109651-13109651 on Build GRCh38
rs747802506 Chr.19: 13109616-13109617 on Build GRCh38
rs763972744 Chr.19: 13109556-13109556 on Build GRCh38
rs756058154 Chr.19: 13109599-13109599 on Build GRCh38
rs554302349 Chr.19: 13109634-13109634 on Build GRCh38
rs756372792 Chr.19: 13109570-13109570 on Build GRCh38
rs780452250 Chr.19: 13109520-13109520 on Build GRCh38
rs774163132 Chr.19: 13109629-13109629 on Build GRCh38
rs572800187 Chr.19: 13109687-13109687 on Build GRCh38
rs574964671 Chr.19: 13109603-13109603 on Build GRCh38
rs756824789 Chr.19: 13109600-13109600 on Build GRCh38
rs367729363 Chr.19: 13109662-13109662 on Build GRCh38
rs757464565 Chr.19: 13109511-13109511 on Build GRCh38
rs769080362 Chr.19: 13109528-13109528 on Build GRCh38
rs376175486 Chr.19: 13109516-13109516 on Build GRCh38
rs772025326 Chr.19: 13109697-13109697 on Build GRCh38
rs746460327 Chr.19: 13109596-13109596 on Build GRCh38
rs149382637 Chr.19: 13109587-13109587 on Build GRCh38
rs768583406 Chr.19: 13109620-13109620 on Build GRCh38
rs540153320 Chr.19: 13109690-13109690 on Build GRCh38
rs932030687 Chr.19: 13109678-13109678 on Build GRCh38
rs1028012377 Chr.19: 13109527-13109527 on Build GRCh38
rs755782799 Chr.19: 13109680-13109680 on Build GRCh38
rs771721929 Chr.19: 13109686-13109686 on Build GRCh38
rs371765162 Chr.19: 13109540-13109540 on Build GRCh38
rs374806942 Chr.19: 13109563-13109563 on Build GRCh38
rs769611083 Chr.19: 13109612-13109612 on Build GRCh38
rs1001975950 Chr.19: 13109597-13109597 on Build GRCh38
rs1009240169 Chr.19: 13109509-13109509 on Build GRCh38
rs868782885 Chr.19: 13109639-13109639 on Build GRCh38
rs780818695 Chr.19: 13109667-13109667 on Build GRCh38
rs201525408 Chr.19: 13109613-13109613 on Build GRCh38
rs761864491 Chr.19: 13109631-13109631 on Build GRCh38
rs1007674574 Chr.19: 13109592-13109592 on Build GRCh38
rs760757803 Chr.19: 13109700-13109700 on Build GRCh38
rs201679777 Chr.19: 13109604-13109604 on Build GRCh38
rs755464364 Chr.19: 13109586-13109586 on Build GRCh38
rs751634698 Chr.19: 13109506-13109506 on Build GRCh38

View Full Product Details