Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116252357-116252582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TES
Entrez Gene ID
26136
Gene Name
testin LIM domain protein
Gene Aliases
TESS, TESS-2
UniGene
Hs.592286
Gene Chromosome Location
Chr.7: 116210493-116258786 on Build GRCh38
Transcripts
NM_015641.3 NM_152829.2 XM_005250258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153772 c.983G>A p.C328Y Chr.7: 116252382-116252382 on Build GRCh38
5345244 c.1063A>T p.K355* Chr.7: 116252462-116252462 on Build GRCh38
4153773 c.983G>A p.C328Y Chr.7: 116252382-116252382 on Build GRCh38
6346168 c.992G>T p.C331F Chr.7: 116252391-116252391 on Build GRCh38
1683398 c.1057_1058insT p.Y353fs*26 Chr.7: 116252456-116252457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757054863 Chr.7: 116252484-116252484 on Build GRCh38
rs201414558 Chr.7: 116252483-116252483 on Build GRCh38
rs866973315 Chr.7: 116252553-116252553 on Build GRCh38
rs775826669 Chr.7: 116252427-116252427 on Build GRCh38
rs762089523 Chr.7: 116252440-116252440 on Build GRCh38
rs376410514 Chr.7: 116252495-116252495 on Build GRCh38
rs777295376 Chr.7: 116252470-116252470 on Build GRCh38
rs776719173 Chr.7: 116252420-116252420 on Build GRCh38
rs770830785 Chr.7: 116252419-116252419 on Build GRCh38
rs758104412 Chr.7: 116252413-116252413 on Build GRCh38
rs774769804 Chr.7: 116252417-116252417 on Build GRCh38
rs988691762 Chr.7: 116252450-116252450 on Build GRCh38
rs769975633 Chr.7: 116252490-116252490 on Build GRCh38
rs199602780 Chr.7: 116252508-116252508 on Build GRCh38
rs747836673 Chr.7: 116252411-116252411 on Build GRCh38
rs201126871 Chr.7: 116252458-116252458 on Build GRCh38
rs745838338 Chr.7: 116252489-116252489 on Build GRCh38
rs143000022 Chr.7: 116252441-116252441 on Build GRCh38
rs908004475 Chr.7: 116252549-116252549 on Build GRCh38
rs117301136 Chr.7: 116252541-116252541 on Build GRCh38
rs752185192 Chr.7: 116252546-116252546 on Build GRCh38
rs770062836 Chr.7: 116252425-116252425 on Build GRCh38
rs758028990 Chr.7: 116252469-116252469 on Build GRCh38
rs774742015 Chr.7: 116252432-116252432 on Build GRCh38
rs764249068 Chr.7: 116252431-116252431 on Build GRCh38
rs200693855 Chr.7: 116252515-116252515 on Build GRCh38
rs778525930 Chr.7: 116252387-116252387 on Build GRCh38
rs199751410 Chr.7: 116252497-116252497 on Build GRCh38
rs202195153 Chr.7: 116252467-116252467 on Build GRCh38
rs762225606 Chr.7: 116252503-116252503 on Build GRCh38
rs764774410 Chr.7: 116252461-116252461 on Build GRCh38
rs564502876 Chr.7: 116252428-116252428 on Build GRCh38
rs200289796 Chr.7: 116252468-116252468 on Build GRCh38
rs200286950 Chr.7: 116252498-116252498 on Build GRCh38
rs781268772 Chr.7: 116252487-116252487 on Build GRCh38
rs763288676 Chr.7: 116252430-116252430 on Build GRCh38
rs151087375 Chr.7: 116252460-116252460 on Build GRCh38
rs201149646 Chr.7: 116252499-116252499 on Build GRCh38
rs1049291119 Chr.7: 116252542-116252542 on Build GRCh38
rs780236160 Chr.7: 116252491-116252491 on Build GRCh38
rs772494465 Chr.7: 116252509-116252509 on Build GRCh38
rs749363435 Chr.7: 116252494-116252494 on Build GRCh38
rs750988155 Chr.7: 116252459-116252459 on Build GRCh38
rs142549105 Chr.7: 116252471-116252471 on Build GRCh38
rs761119871 Chr.7: 116252535-116252535 on Build GRCh38
rs554017721 Chr.7: 116252418-116252418 on Build GRCh38
rs539484837 Chr.7: 116252527-116252527 on Build GRCh38
rs1003349030 Chr.7: 116252442-116252442 on Build GRCh38

View Full Product Details