Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 188140463-188140736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIML1
Entrez Gene ID
339976
Gene Name
tripartite motif family like 1
Gene Aliases
RNF209
UniGene
Hs.348618
Gene Chromosome Location
Chr.4: 188138189-188150768 on Build GRCh38
Transcripts
NM_178556.3 XM_006714207.3 XM_011531919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011724 c.478G>A p.E160K Chr.4: 188140597-188140597 on Build GRCh38
6512758 c.417C>A p.L139L Chr.4: 188140536-188140536 on Build GRCh38
6497320 c.460C>A p.Q154K Chr.4: 188140579-188140579 on Build GRCh38
1540185 c.439C>T p.R147C Chr.4: 188140558-188140558 on Build GRCh38
1230426 c.434T>G p.L145R Chr.4: 188140553-188140553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752025162 Chr.4: 188140559-188140559 on Build GRCh38
rs201368764 Chr.4: 188140509-188140509 on Build GRCh38
rs946332481 Chr.4: 188140555-188140555 on Build GRCh38
rs767151289 Chr.4: 188140598-188140600 on Build GRCh38
rs777178757 Chr.4: 188140626-188140626 on Build GRCh38
rs373412379 Chr.4: 188140677-188140677 on Build GRCh38
rs765837635 Chr.4: 188140545-188140545 on Build GRCh38
rs761886564 Chr.4: 188140529-188140529 on Build GRCh38
rs370775791 Chr.4: 188140633-188140633 on Build GRCh38
rs372971866 Chr.4: 188140510-188140510 on Build GRCh38
rs765250470 Chr.4: 188140543-188140543 on Build GRCh38
rs540407052 Chr.4: 188140673-188140673 on Build GRCh38
rs374459667 Chr.4: 188140641-188140641 on Build GRCh38
rs771630304 Chr.4: 188140672-188140672 on Build GRCh38
rs768016565 Chr.4: 188140675-188140675 on Build GRCh38
rs376112585 Chr.4: 188140627-188140627 on Build GRCh38
rs897736760 Chr.4: 188140685-188140685 on Build GRCh38
rs753190119 Chr.4: 188140590-188140590 on Build GRCh38
rs111987767 Chr.4: 188140664-188140664 on Build GRCh38
rs965533022 Chr.4: 188140491-188140491 on Build GRCh38
rs777708257 Chr.4: 188140502-188140502 on Build GRCh38
rs556207095 Chr.4: 188140542-188140542 on Build GRCh38
rs944880324 Chr.4: 188140517-188140517 on Build GRCh38
rs62354297 Chr.4: 188140669-188140669 on Build GRCh38
rs767208782 Chr.4: 188140575-188140575 on Build GRCh38
rs571253883 Chr.4: 188140701-188140702 on Build GRCh38
rs745730600 Chr.4: 188140636-188140636 on Build GRCh38
rs553977858 Chr.4: 188140496-188140496 on Build GRCh38
rs754669930 Chr.4: 188140569-188140569 on Build GRCh38
rs773723882 Chr.4: 188140549-188140549 on Build GRCh38
rs138341929 Chr.4: 188140702-188140703 on Build GRCh38
rs775056117 Chr.4: 188140616-188140617 on Build GRCh38
rs186485922 Chr.4: 188140707-188140707 on Build GRCh38
rs1034521816 Chr.4: 188140646-188140646 on Build GRCh38
rs766822406 Chr.4: 188140558-188140558 on Build GRCh38
rs781422167 Chr.4: 188140651-188140651 on Build GRCh38
rs778667196 Chr.4: 188140634-188140634 on Build GRCh38
rs776569510 Chr.4: 188140525-188140525 on Build GRCh38
rs755601791 Chr.4: 188140595-188140595 on Build GRCh38
rs904200407 Chr.4: 188140599-188140599 on Build GRCh38
rs763404758 Chr.4: 188140550-188140550 on Build GRCh38
rs752301460 Chr.4: 188140581-188140581 on Build GRCh38
rs796769014 Chr.4: 188140702-188140702 on Build GRCh38
rs748344710 Chr.4: 188140654-188140654 on Build GRCh38
rs539685958 Chr.4: 188140511-188140511 on Build GRCh38
rs1043054135 Chr.4: 188140567-188140567 on Build GRCh38
rs182772435 Chr.4: 188140671-188140671 on Build GRCh38
rs113738610 Chr.4: 188140521-188140522 on Build GRCh38
rs946951763 Chr.4: 188140561-188140561 on Build GRCh38

View Full Product Details