Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49168392-49168653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM4
Entrez Gene ID
54795
Gene Name
transient receptor potential cation channel subfamily M member 4
Gene Aliases
LTrpC4, PFHB1B, TRPM4B, hTRPM4
UniGene
Hs.467101
Gene Chromosome Location
Chr.19: 49157741-49211841 on Build GRCh38
Transcripts
NM_001195227.1 NM_001321281.1 NM_001321282.1 NM_001321283.1 NM_001321285.1 NM_017636.3 XM_005259017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712581 c.617C>T p.S206L Chr.19: 49168557-49168557 on Build GRCh38
3536765 c.668T>G p.F223C Chr.19: 49168608-49168608 on Build GRCh38
4436609 c.660G>T p.G220G Chr.19: 49168600-49168600 on Build GRCh38
5540913 c.699C>T p.F233F Chr.19: 49168639-49168639 on Build GRCh38
6151974 c.651G>T p.P217P Chr.19: 49168591-49168591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs730880236 Chr.19: 49168618-49168618 on Build GRCh38
rs373566236 Chr.19: 49168525-49168525 on Build GRCh38
rs148499675 Chr.19: 49168611-49168611 on Build GRCh38
rs779097169 Chr.19: 49168429-49168429 on Build GRCh38
rs1040381781 Chr.19: 49168484-49168484 on Build GRCh38
rs199597602 Chr.19: 49168523-49168523 on Build GRCh38
rs1023173897 Chr.19: 49168522-49168522 on Build GRCh38
rs777041419 Chr.19: 49168538-49168538 on Build GRCh38
rs1006381109 Chr.19: 49168516-49168516 on Build GRCh38
rs747502300 Chr.19: 49168592-49168592 on Build GRCh38
rs376048499 Chr.19: 49168492-49168492 on Build GRCh38
rs758431744 Chr.19: 49168526-49168526 on Build GRCh38
rs757555185 Chr.19: 49168515-49168515 on Build GRCh38
rs866558752 Chr.19: 49168422-49168422 on Build GRCh38
rs556601275 Chr.19: 49168529-49168529 on Build GRCh38
rs371319252 Chr.19: 49168583-49168583 on Build GRCh38
rs748931206 Chr.19: 49168630-49168630 on Build GRCh38
rs558212476 Chr.19: 49168629-49168629 on Build GRCh38
rs759593186 Chr.19: 49168574-49168574 on Build GRCh38
rs901893809 Chr.19: 49168485-49168485 on Build GRCh38
rs377694905 Chr.19: 49168504-49168504 on Build GRCh38
rs774397450 Chr.19: 49168567-49168567 on Build GRCh38
rs551910601 Chr.19: 49168469-49168469 on Build GRCh38
rs570618364 Chr.19: 49168507-49168507 on Build GRCh38
rs190211117 Chr.19: 49168545-49168545 on Build GRCh38
rs772702533 Chr.19: 49168443-49168443 on Build GRCh38
rs149860526 Chr.19: 49168580-49168580 on Build GRCh38
rs761505972 Chr.19: 49168459-49168459 on Build GRCh38
rs373953725 Chr.19: 49168597-49168597 on Build GRCh38
rs773243853 Chr.19: 49168448-49168448 on Build GRCh38
rs762377900 Chr.19: 49168503-49168503 on Build GRCh38
rs758473371 Chr.19: 49168591-49168591 on Build GRCh38
rs750021579 Chr.19: 49168470-49168470 on Build GRCh38
rs776237070 Chr.19: 49168458-49168458 on Build GRCh38
rs866131806 Chr.19: 49168420-49168420 on Build GRCh38
rs775865025 Chr.19: 49168449-49168449 on Build GRCh38
rs750644014 Chr.19: 49168590-49168590 on Build GRCh38
rs762472792 Chr.19: 49168563-49168563 on Build GRCh38
rs769040648 Chr.19: 49168457-49168457 on Build GRCh38
rs113159745 Chr.19: 49168514-49168514 on Build GRCh38
rs768987697 Chr.19: 49168531-49168531 on Build GRCh38
rs201198341 Chr.19: 49168636-49168636 on Build GRCh38
rs145878044 Chr.19: 49168581-49168581 on Build GRCh38
rs755866022 Chr.19: 49168582-49168582 on Build GRCh38
rs142788545 Chr.19: 49168558-49168558 on Build GRCh38
rs866398035 Chr.19: 49168603-49168603 on Build GRCh38
rs376782481 Chr.19: 49168557-49168557 on Build GRCh38
rs746345680 Chr.19: 49168441-49168441 on Build GRCh38
rs770073838 Chr.19: 49168627-49168627 on Build GRCh38
rs546529855 Chr.19: 49168617-49168617 on Build GRCh38
rs747796161 Chr.19: 49168548-49168548 on Build GRCh38
rs781170686 Chr.19: 49168508-49168508 on Build GRCh38
rs183306159 Chr.19: 49168575-49168575 on Build GRCh38
rs770203558 Chr.19: 49168561-49168561 on Build GRCh38
rs943322712 Chr.19: 49168467-49168467 on Build GRCh38
rs911181193 Chr.19: 49168438-49168438 on Build GRCh38
rs765832900 Chr.19: 49168566-49168566 on Build GRCh38

View Full Product Details