Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 102438399-102438660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZPLD1
Entrez Gene ID
131368
Gene Name
zona pellucida like domain containing 1
Gene Aliases
-
UniGene
Hs.352213
Gene Chromosome Location
Chr.3: 102385144-102479841 on Build GRCh38
Transcripts
NM_175056.1 XM_017005703.1 XM_017005704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333419 c.97G>T p.G33W Chr.3: 102438536-102438536 on Build GRCh38
4447638 c.41-3C>T p.? Chr.3: 102438477-102438477 on Build GRCh38
4188126 c.62G>T p.W21L Chr.3: 102438501-102438501 on Build GRCh38
6834139 c.114C>T p.N38N Chr.3: 102438553-102438553 on Build GRCh38
4929311 c.104C>T p.A35V Chr.3: 102438543-102438543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898824520 Chr.3: 102438453-102438453 on Build GRCh38
rs534712898 Chr.3: 102438613-102438613 on Build GRCh38
rs189843915 Chr.3: 102438629-102438629 on Build GRCh38
rs529972484 Chr.3: 102438451-102438451 on Build GRCh38
rs886154571 Chr.3: 102438466-102438466 on Build GRCh38
rs753480764 Chr.3: 102438574-102438574 on Build GRCh38
rs11340850 Chr.3: 102438440-102438440 on Build GRCh38
rs6784362 Chr.3: 102438521-102438521 on Build GRCh38
rs779045061 Chr.3: 102438438-102438438 on Build GRCh38
rs561217700 Chr.3: 102438427-102438427 on Build GRCh38
rs755138135 Chr.3: 102438597-102438597 on Build GRCh38
rs1007825928 Chr.3: 102438594-102438594 on Build GRCh38
rs6784389 Chr.3: 102438573-102438573 on Build GRCh38
rs868030139 Chr.3: 102438593-102438593 on Build GRCh38
rs770543242 Chr.3: 102438542-102438542 on Build GRCh38
rs772565788 Chr.3: 102438528-102438528 on Build GRCh38
rs779114687 Chr.3: 102438604-102438604 on Build GRCh38
rs543688254 Chr.3: 102438464-102438464 on Build GRCh38
rs749477019 Chr.3: 102438500-102438500 on Build GRCh38
rs139525039 Chr.3: 102438590-102438590 on Build GRCh38
rs776311841 Chr.3: 102438545-102438545 on Build GRCh38
rs1012492061 Chr.3: 102438540-102438540 on Build GRCh38
rs775507307 Chr.3: 102438610-102438611 on Build GRCh38
rs765981966 Chr.3: 102438436-102438436 on Build GRCh38
rs751425757 Chr.3: 102438559-102438559 on Build GRCh38
rs751578898 Chr.3: 102438468-102438468 on Build GRCh38
rs199954809 Chr.3: 102438534-102438534 on Build GRCh38
rs759215236 Chr.3: 102438546-102438546 on Build GRCh38
rs574774931 Chr.3: 102438609-102438609 on Build GRCh38
rs761304064 Chr.3: 102438535-102438535 on Build GRCh38
rs112773486 Chr.3: 102438493-102438493 on Build GRCh38
rs764994042 Chr.3: 102438550-102438550 on Build GRCh38
rs757485308 Chr.3: 102438429-102438429 on Build GRCh38
rs747508648 Chr.3: 102438554-102438554 on Build GRCh38
rs768901721 Chr.3: 102438503-102438503 on Build GRCh38
rs145035940 Chr.3: 102438462-102438462 on Build GRCh38
rs762649112 Chr.3: 102438553-102438553 on Build GRCh38
rs370088641 Chr.3: 102438596-102438596 on Build GRCh38
rs759311326 Chr.3: 102438446-102438446 on Build GRCh38
rs144383721 Chr.3: 102438496-102438496 on Build GRCh38
rs774216250 Chr.3: 102438556-102438556 on Build GRCh38
rs757254249 Chr.3: 102438571-102438571 on Build GRCh38
rs760109568 Chr.3: 102438432-102438432 on Build GRCh38
rs777574806 Chr.3: 102438465-102438465 on Build GRCh38
rs752472400 Chr.3: 102438552-102438552 on Build GRCh38
rs910451277 Chr.3: 102438537-102438537 on Build GRCh38
rs756130969 Chr.3: 102438484-102438484 on Build GRCh38
rs1035183487 Chr.3: 102438529-102438529 on Build GRCh38
rs537289075 Chr.3: 102438551-102438551 on Build GRCh38

View Full Product Details