Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58137880-58138131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 NM_001320173.1 XM_011536471.2 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5705737 c.1313T>C p.V438A Chr.14: 58138046-58138046 on Build GRCh38
4823024 c.1268A>T p.E423V Chr.14: 58138091-58138091 on Build GRCh38
7121591 c.1287T>C p.D429D Chr.14: 58138072-58138072 on Build GRCh38
1493264 c.1369+2T>A p.? Chr.14: 58137988-58137988 on Build GRCh38
433172 c.1229C>T p.S410F Chr.14: 58138130-58138130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370881117 Chr.14: 58138019-58138019 on Build GRCh38
rs367892858 Chr.14: 58137973-58137973 on Build GRCh38
rs747499015 Chr.14: 58137976-58137979 on Build GRCh38
rs912772440 Chr.14: 58137952-58137952 on Build GRCh38
rs921588026 Chr.14: 58138047-58138047 on Build GRCh38
rs1052643898 Chr.14: 58137917-58137917 on Build GRCh38
rs944309905 Chr.14: 58137977-58137977 on Build GRCh38
rs142858496 Chr.14: 58138103-58138103 on Build GRCh38
rs768503569 Chr.14: 58138101-58138101 on Build GRCh38
rs183669375 Chr.14: 58138075-58138075 on Build GRCh38
rs566072175 Chr.14: 58138099-58138099 on Build GRCh38
rs768534999 Chr.14: 58137957-58137957 on Build GRCh38
rs1016990214 Chr.14: 58138051-58138051 on Build GRCh38
rs764857280 Chr.14: 58137983-58137983 on Build GRCh38
rs775187852 Chr.14: 58137947-58137947 on Build GRCh38
rs764438987 Chr.14: 58138053-58138053 on Build GRCh38
rs756637157 Chr.14: 58138043-58138043 on Build GRCh38
rs371735875 Chr.14: 58137979-58137979 on Build GRCh38
rs933528486 Chr.14: 58137931-58137931 on Build GRCh38
rs529338598 Chr.14: 58137949-58137949 on Build GRCh38
rs180703797 Chr.14: 58138050-58138050 on Build GRCh38
rs901977663 Chr.14: 58137928-58137928 on Build GRCh38
rs746848331 Chr.14: 58138094-58138094 on Build GRCh38
rs200905995 Chr.14: 58138106-58138106 on Build GRCh38
rs762729288 Chr.14: 58137992-58137992 on Build GRCh38
rs1054208208 Chr.14: 58138082-58138082 on Build GRCh38
rs778994236 Chr.14: 58138071-58138071 on Build GRCh38
rs769342602 Chr.14: 58138106-58138140 on Build GRCh38
rs566768348 Chr.14: 58138060-58138060 on Build GRCh38
rs762919140 Chr.14: 58137963-58137966 on Build GRCh38
rs960527801 Chr.14: 58138086-58138086 on Build GRCh38
rs61745856 Chr.14: 58138093-58138093 on Build GRCh38
rs137871178 Chr.14: 58138012-58138012 on Build GRCh38
rs757961560 Chr.14: 58137967-58137968 on Build GRCh38

View Full Product Details