Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51302895-51303126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39A
Entrez Gene ID
22996
Gene Name
tetratricopeptide repeat domain 39A
Gene Aliases
C1orf34, DEME-6
UniGene
Hs.112949
Gene Chromosome Location
Chr.1: 51287258-51345116 on Build GRCh38
Transcripts
NM_001080494.3 NM_001144832.2 NM_001297662.1 NM_001297663.1 NM_001297664.1 NM_001297665.1 XM_005270644.2 XM_006710471.2 XM_011541048.2 XM_011541049.2 XM_011541050.2 XM_011541051.2 XM_011541052.2 XM_011541053.2 XM_011541054.2 XM_017000715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5460774 c.652T>C p.C218R Chr.1: 51303114-51303114 on Build GRCh38
2192356 c.658C>A p.H220N Chr.1: 51303108-51303108 on Build GRCh38
3400864 c.865G>A p.V289M Chr.1: 51303090-51303090 on Build GRCh38
3400865 c.676G>A p.V226M Chr.1: 51303090-51303090 on Build GRCh38
2192357 c.835C>A p.H279N Chr.1: 51303108-51303108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765602735 Chr.1: 51303034-51303034 on Build GRCh38
rs1020776806 Chr.1: 51303016-51303016 on Build GRCh38
rs576401430 Chr.1: 51303007-51303007 on Build GRCh38
rs778731397 Chr.1: 51303068-51303068 on Build GRCh38
rs1019654639 Chr.1: 51302984-51302984 on Build GRCh38
rs186731507 Chr.1: 51303048-51303048 on Build GRCh38
rs182775460 Chr.1: 51303045-51303045 on Build GRCh38
rs747162456 Chr.1: 51303094-51303094 on Build GRCh38
rs563904757 Chr.1: 51303106-51303106 on Build GRCh38
rs771991689 Chr.1: 51303084-51303084 on Build GRCh38
rs749886189 Chr.1: 51303047-51303047 on Build GRCh38
rs535925697 Chr.1: 51302922-51302922 on Build GRCh38
rs974512063 Chr.1: 51302993-51302993 on Build GRCh38
rs1008202314 Chr.1: 51302958-51302959 on Build GRCh38
rs191762418 Chr.1: 51303008-51303008 on Build GRCh38
rs745861337 Chr.1: 51303038-51303038 on Build GRCh38
rs539606227 Chr.1: 51302945-51302945 on Build GRCh38
rs191483055 Chr.1: 51303089-51303089 on Build GRCh38
rs756712354 Chr.1: 51303073-51303073 on Build GRCh38
rs117188633 Chr.1: 51302924-51302924 on Build GRCh38
rs910438329 Chr.1: 51303023-51303023 on Build GRCh38
rs201005287 Chr.1: 51303052-51303052 on Build GRCh38
rs748060694 Chr.1: 51303077-51303077 on Build GRCh38
rs753073744 Chr.1: 51303057-51303057 on Build GRCh38
rs777433920 Chr.1: 51302940-51302940 on Build GRCh38
rs896729450 Chr.1: 51303071-51303071 on Build GRCh38
rs780543256 Chr.1: 51303022-51303022 on Build GRCh38
rs1014203096 Chr.1: 51303104-51303104 on Build GRCh38
rs558569398 Chr.1: 51303067-51303067 on Build GRCh38
rs142217948 Chr.1: 51303005-51303005 on Build GRCh38
rs369769209 Chr.1: 51303039-51303039 on Build GRCh38
rs753520447 Chr.1: 51303066-51303066 on Build GRCh38
rs566182685 Chr.1: 51302943-51302943 on Build GRCh38
rs777724015 Chr.1: 51303090-51303090 on Build GRCh38
rs74356055 Chr.1: 51303044-51303044 on Build GRCh38

View Full Product Details